This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06829820_cn
See other AK8 CNV Assays ›
Gene Symbol
AK8
Assay Reference Genome
Location

Chr.9:132730847 on build GRCh38
Cytoband
9q34.13
Assay ID Hs06829820_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

158067

Gene Name:

adenylate kinase 8

Gene Aliases:

AK 8, C9orf98, DDX31

Location:

Chr.9:132725574-132878811 on Build GRCh38

Assay Gene Location:

Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AK8 NM_001317958.1 NP_001304887.1
NM_001317959.1 NP_001304888.1
NM_152572.2 NP_689785.1
XM_005272169.2 XP_005272226.1
XM_006716965.2 XP_006717028.1
XM_011518278.2 XP_011516580.1
XM_017014308.1 XP_016869797.1
AK057266.1 BAB71402.1
AK093446.1 BAC04168.1
AK292186.1
BC034776.1 AAH34776.1
BC050576.1 AAH50576.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1051206 Chr.9:132723216 - 132863839 on Build GRCh38 Gain AK8
esv2739138 Chr.9:132727483 - 135521553 on Build GRCh38 Deletion RPL7A LINC00094 LOC101928525 MIR3689C SNORD141A SPACA9 LOC105376306 FAM163B DBH FCN1 WDR5 MIR3689E ADAMTSL2 PPP1R26 SARDH RALGDS RXRA GTF3C5 LOC101448202 MED22 MIR3689D1 MIR4669 SLC2A6 LOC401557 LOC100996574 BRD3 COL5A1 SNORD36A C9orf62 MIR3689D2 LOC100130548 MIR3689A OBP2B SURF1 DBH-AS1 MIR3689B ABO SNORD36B CELP OLFM1 GBGT1 REXO4 FCN2 ADAMTS13 STKLD1 TSC1 LOC101928193 MIR6877 COL5A1-AS1 C9orf116 AK8 SNORD36C LCN1 CACFD1 SURF4 SNORD24 TMEM8C MIR548AW RNU6ATAC VAV2 PPP1R26-AS1 SURF2 SURF6 CEL MIR3689F GFI1B MRPS2
dgv1513e214 Chr.9:132713668 - 132865324 on Build GRCh38 Gain AK8

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-vvlg5:80/100.66.78.195:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline