This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06864046_cn
See other AK8 CNV Assays ›
Gene Symbol
AK8
Assay Reference Genome
Location

Chr.9:132730245 on build GRCh38
Cytoband
9q34.13
Assay ID Hs06864046_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

158067

Gene Name:

adenylate kinase 8

Gene Aliases:

AK 8, C9orf98, DDX31

Location:

Chr.9:132725574-132878811 on Build GRCh38

Assay Gene Location:

Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AK8 NM_001317958.1 NP_001304887.1
NM_001317959.1 NP_001304888.1
NM_152572.2 NP_689785.1
XM_005272169.2 XP_005272226.1
XM_006716965.2 XP_006717028.1
XM_011518278.2 XP_011516580.1
XM_017014308.1 XP_016869797.1
AK057266.1 BAB71402.1
AK093446.1 BAC04168.1
AK292186.1
BC034776.1 AAH34776.1
BC050576.1 AAH50576.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1051206 Chr.9:132723216 - 132863839 on Build GRCh38 Gain AK8
esv2739138 Chr.9:132727483 - 135521553 on Build GRCh38 Deletion STKLD1 C9orf62 ADAMTS13 DBH-AS1 SNORD141A C9orf116 SNORD36B MIR6877 MIR3689B SPACA9 WDR5 MIR4669 RNU6ATAC SARDH MIR3689E SURF4 SNORD36A BRD3 MIR3689D2 SURF6 MIR548AW ADAMTSL2 SLC2A6 MIR3689C LOC401557 FCN2 LOC100996574 TSC1 AK8 VAV2 MED22 FCN1 LOC105376306 PPP1R26-AS1 SNORD36C MIR3689A GFI1B FAM163B MIR3689D1 SURF2 MIR3689F SNORD24 LCN1 LOC101928525 GBGT1 LOC101928193 OLFM1 DBH PPP1R26 ABO LINC00094 RPL7A REXO4 CELP COL5A1 SURF1 GTF3C5 MRPS2 OBP2B RXRA LOC100130548 RALGDS CACFD1 COL5A1-AS1 LOC101448202 CEL TMEM8C
dgv1513e214 Chr.9:132713668 - 132865324 on Build GRCh38 Gain AK8

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-jmhj8:80/100.66.74.142:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline