This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06886913_cn
See other AK8 CNV Assays ›
Gene Symbol
AK8
Assay Reference Genome
Location

Chr.9:132732978 on build GRCh38
Cytoband
9q34.13
Assay ID Hs06886913_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

158067

Gene Name:

adenylate kinase 8

Gene Aliases:

AK 8, C9orf98, DDX31

Location:

Chr.9:132725574-132878811 on Build GRCh38

Assay Gene Location:

Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AK8 NM_001317958.1 NP_001304887.1
NM_001317959.1 NP_001304888.1
NM_152572.2 NP_689785.1
XM_005272169.2 XP_005272226.1
XM_006716965.2 XP_006717028.1
XM_011518278.2 XP_011516580.1
XM_017014308.1 XP_016869797.1
AK057266.1 BAB71402.1
AK093446.1 BAC04168.1
AK292186.1
BC034776.1 AAH34776.1
BC050576.1 AAH50576.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1051206 Chr.9:132723216 - 132863839 on Build GRCh38 Gain AK8
esv2739138 Chr.9:132727483 - 135521553 on Build GRCh38 Deletion DBH MIR3689D2 COL5A1 GFI1B FAM163B SNORD36A AK8 LOC101928525 OLFM1 ABO CELP SARDH RPL7A LOC101448202 SURF4 LOC100130548 CEL BRD3 SNORD36B PPP1R26 SNORD36C MIR548AW RNU6ATAC LINC00094 MED22 SNORD24 LOC100996574 FCN1 WDR5 LOC105376306 STKLD1 SURF6 REXO4 C9orf116 SNORD141A ADAMTSL2 PPP1R26-AS1 GBGT1 FCN2 SURF1 SPACA9 MIR3689F MIR4669 MRPS2 MIR3689D1 TMEM8C TSC1 MIR3689B OBP2B VAV2 RALGDS COL5A1-AS1 ADAMTS13 SURF2 CACFD1 LOC101928193 MIR3689A MIR3689C DBH-AS1 C9orf62 LCN1 GTF3C5 LOC401557 MIR6877 SLC2A6 RXRA MIR3689E
dgv1513e214 Chr.9:132713668 - 132865324 on Build GRCh38 Gain AK8

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-vvlg5:80/100.66.78.195:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline