This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06909712_cn
See other LOC105375975 CNV Assays ›
Gene Symbol
LOC105375975
Assay Reference Genome
Location

Chr.9:11614805 on build GRCh38
Cytoband
9p23
Assay ID Hs06909712_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

105375975

Gene Name:

histone-lysine N-methyltransferase SETMAR-like

Gene Aliases:

-

Location:

Chr.9:11610222-11618749 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC105375975 XM_011518088.2 XP_011516390.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2752276 Chr.9:11224627 - 11958106 on Build GRCh38 Loss LOC105375975
esv2422486 Chr.9:11512989 - 11675590 on Build GRCh38 Deletion LOC105375975
nsv1028493 Chr.9:11607030 - 11622644 on Build GRCh38 Gain LOC105375975
esv2761283 Chr.9:11523636 - 12014548 on Build GRCh38 Loss LOC105375975
nsv613433 Chr.9:11606348 - 11716862 on Build GRCh38 Gain LOC105375975
nsv1017027 Chr.9:11575785 - 11799681 on Build GRCh38 Gain LOC105375975
nsv613420 Chr.9:11200588 - 11619839 on Build GRCh38 Loss LOC105375975
nsv1034601 Chr.9:11596157 - 11623397 on Build GRCh38 Loss LOC105375975
nsv8415 Chr.9:11173118 - 11631933 on Build GRCh38 Loss LOC105375975
esv2752278 Chr.9:11600500 - 11701727 on Build GRCh38 Loss LOC105375975
esv3619616 Chr.9:11600555 - 11733459 on Build GRCh38 Loss LOC105375975
nsv466186 Chr.9:11453474 - 11645495 on Build GRCh38 Loss LOC105375975
dgv7411n100 Chr.9:11459329 - 11631105 on Build GRCh38 Loss LOC105375975
dgv7412n100 Chr.9:11582831 - 11934365 on Build GRCh38 Loss LOC105375975
esv3619599 Chr.9:11347309 - 11623122 on Build GRCh38 Loss LOC105375975
esv3619615 Chr.9:11595197 - 11623463 on Build GRCh38 Loss LOC105375975
dgv7413n100 Chr.9:11600297 - 11841788 on Build GRCh38 Loss LOC105375975
esv3891582 Chr.9:11511860 - 11787410 on Build GRCh38 Loss LOC105375975

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

viral or transposable element protein

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0