This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06922521_cn
See other AK8 CNV Assays ›
Gene Symbol
AK8
Assay Reference Genome
Location

Chr.9:132733704 on build GRCh38
Cytoband
9q34.13
Assay ID Hs06922521_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

158067

Gene Name:

adenylate kinase 8

Gene Aliases:

AK 8, C9orf98, DDX31

Location:

Chr.9:132725574-132878811 on Build GRCh38

Assay Gene Location:

Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AK8 NM_001317958.1 NP_001304887.1
NM_001317959.1 NP_001304888.1
NM_152572.2 NP_689785.1
XM_005272169.2 XP_005272226.1
XM_006716965.2 XP_006717028.1
XM_011518278.2 XP_011516580.1
XM_017014308.1 XP_016869797.1
AK057266.1 BAB71402.1
AK093446.1 BAC04168.1
AK292186.1
BC034776.1 AAH34776.1
BC050576.1 AAH50576.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1051206 Chr.9:132723216 - 132863839 on Build GRCh38 Gain AK8
esv2739138 Chr.9:132727483 - 135521553 on Build GRCh38 Deletion DBH-AS1 SNORD141A RPL7A SLC2A6 SNORD36B COL5A1-AS1 MIR3689C MIR3689E COL5A1 LCN1 MIR548AW MIR6877 SPACA9 TMEM8C C9orf62 MIR3689D2 SURF4 SNORD36C MIR3689F ABO SURF1 LINC00094 CELP SURF6 GBGT1 FAM163B LOC100130548 MED22 LOC101928193 SNORD36A MIR3689A LOC101928525 RALGDS OBP2B SNORD24 MRPS2 LOC100996574 MIR4669 LOC401557 FCN2 AK8 PPP1R26 VAV2 GTF3C5 OLFM1 DBH BRD3 RNU6ATAC CACFD1 CEL TSC1 PPP1R26-AS1 MIR3689B SARDH RXRA SURF2 GFI1B LOC101448202 LOC105376306 ADAMTSL2 C9orf116 WDR5 ADAMTS13 REXO4 MIR3689D1 FCN1 STKLD1
nsv615574 Chr.9:132733601 - 132751450 on Build GRCh38 Loss AK8
dgv1513e214 Chr.9:132713668 - 132865324 on Build GRCh38 Gain AK8

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-v472f:80/100.66.78.195:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline