This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06976039_cn
See other TDG CNV Assays ›
Gene Symbol
TDG
Assay Reference Genome
Location

Chr.12:103972996 on build GRCh38
Cytoband
12q23.3
Assay ID Hs06976039_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

6996

Gene Name:

thymine DNA glycosylase

Gene Aliases:

hTDG

Location:

Chr.12:103965815-103988878 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
TDG NM_003211.4 NP_003202.3
XM_005269125.1 XP_005269182.1
XM_011538714.1 XP_011537016.1
XM_011538715.1 XP_011537017.1
AK299832.1
AK303631.1
AK309792.1
AK311673.1
AK313234.1
AL710869.1
BC019925.1
BC037557.1 AAH37557.1
BC104477.1 AAI04478.1
CB047650.1
DB162106.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv560039 Chr.12:103783890 - 104061562 on Build GRCh38 Gain TTC41P NT5DC3 C12orf73 MIR3652 GLT8D2 HSP90B1 TDG
esv2746302 Chr.12:103966022 - 103979831 on Build GRCh38 Deletion TDG

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
base-excision repair
base-excision repair, AP site formation
mismatch repair
transcription, DNA-templated
embryo development
covalent chromatin modification
protein sumoylation
negative regulation of protein binding
oxidative DNA demethylation
negative regulation of chromatin binding
regulation of gene expression, epigenetic
depyrimidination
DNA demethylation
RNA polymerase II transcription cofactor activity
damaged DNA binding
double-stranded DNA binding
uracil DNA N-glycosylase activity
protein kinase C binding
protein binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
DNA N-glycosylase activity
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0