This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07001288_cn
See other POLE CNV Assays ›
Gene Symbol
POLE
Assay Reference Genome
Location

Chr.12:132631615 on build GRCh38
Cytoband
12q24.33
Assay ID Hs07001288_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5426

Gene Name:

DNA polymerase epsilon, catalytic subunit

Gene Aliases:

CRCS12, FILS, POLE1

Location:

Chr.12:132623762-132687524 on Build GRCh38

Assay Gene Location:

Within Intron 47
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
POLE NM_006231.3 NP_006222.2
XM_011534795.2 XP_011533097.1
XM_011534797.2 XP_011533099.1
XM_011534798.1 XP_011533100.1
XM_011534802.2 XP_011533104.1
AK025087.1 BAB15063.1
AK093003.1
BC007599.1 AAH07599.1
BC021559.1 AAH21559.1
BC087613.1 AAH87613.1
BX647647.1
L09561.1 AAC19148.1
S60080.1 AAA15448.1
U49356.1 AAA90924.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv561110 Chr.12:132630826 - 132677497 on Build GRCh38 Loss POLE
nsv952512 Chr.12:132192256 - 132711214 on Build GRCh38 Deletion LOC105370092 POLE GALNT9 FBRSL1 PGAM5 MUC8 LOC100130238 PXMP2 P2RX2 LRCOL1 LOC100996573 LOC101928416 MIR6763
esv2422284 Chr.12:132604248 - 132708921 on Build GRCh38 Deletion PXMP2 P2RX2 LRCOL1 POLE
nsv1037074 Chr.12:132613063 - 132801953 on Build GRCh38 Gain PXMP2 P2RX2 GOLGA3 ANKLE2 POLE PGAM5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
telomere maintenance via recombination
DNA synthesis involved in DNA repair
DNA replication
DNA replication initiation
base-excision repair, gap-filling
nucleotide-excision repair, DNA gap filling
embryonic organ development
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
DNA binding
chromatin binding
DNA-directed DNA polymerase activity
zinc ion binding
3'-5' exonuclease activity
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0