This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07015657_cn
See other CD163L1 CNV Assays ›
Gene Symbol
CD163L1
Assay Reference Genome
Location

Chr.12:7342981 on build GRCh38
Cytoband
12p13.31
Assay ID Hs07015657_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

283316

Gene Name:

CD163 molecule like 1

Gene Aliases:

CD163B, M160, SCARI2

Location:

Chr.12:7320510-7444185 on Build GRCh38

Assay Gene Location:

Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD163L1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv448e212 Chr.12:7280402 - 7344654 on Build GRCh38 Gain CD163L1 ACSM4
nsv594 Chr.12:7329134 - 7363501 on Build GRCh38 Deletion CD163L1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline