This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07057692_cn
See other DDX24 CNV Assays ›
Gene Symbol
DDX24
Assay Reference Genome
Location

Chr.14:94078377 on build GRCh38
Cytoband
14q32.12
Assay ID Hs07057692_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57062

Gene Name:

DEAD-box helicase 24

Gene Aliases:

-

Location:

Chr.14:94050922-94081212 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DDX24 NM_020414.3 NP_065147.1
AB209382.1 BAD92619.1
AF214731.1 AAG02169.1
AK025162.1 BAB15079.1
AK294613.1
AK296726.1
AK297232.1
AK304504.1
AL136886.1 CAB66820.1
BC008847.2 AAH08847.1
BC009406.2 AAH09406.1
BC096826.1 AAH96826.1
BX537533.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv34067 Chr.14:94024355 - 94160851 on Build GRCh38 Loss IFI27 IFI27L2 IFI27L1 DDX24 OTUB2
esv3635373 Chr.14:94040151 - 94173989 on Build GRCh38 Gain IFI27 IFI27L2 IFI27L1 DDX24 OTUB2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

RNA secondary structure unwinding
RNA metabolic process
RNA helicase activity
ATP-dependent RNA helicase activity
ATP binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-548c9458fd-zf69l:80/100.66.73.23:80.
git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.0-Offline