This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07062463_cn
See other NIN CNV Assays ›
Gene Symbol
NIN
Assay Reference Genome
Location

Chr.14:50730512 on build GRCh38
Cytoband
14q22.1
Assay ID Hs07062463_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51199

Gene Name:

ninein

Gene Aliases:

SCKL7

Location:

Chr.14:50719763-50831121 on Build GRCh38

Assay Gene Location:

Within Intron 30
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NIN NM_016350.4 NP_057434.4
NM_020921.3 NP_065972.3
NM_182944.2 NP_891989.2
NM_182946.1 NP_891991.1
XM_011536817.1 XP_011535119.1
XM_011536819.2 XP_011535121.1
XM_011536822.1 XP_011535124.1
XM_011536823.2 XP_011535125.1
AB046785.2 BAB13391.2
AF186776.1 AAG17027.1
AF212162.2 AAF23015.2
AF223937.1 AAK00628.1
AF223938.1 AAK00629.1
AF223939.1 AAK00630.1
AF302773.2 AAG33512.2
AK025652.1
AK026083.1
AK027054.1 BAB15640.1
AY027794.1 AAK27375.1
AY027795.1 AAK27376.1
AY027796.1 AAK27377.1
AY736182.1 AAU86898.1
AY736183.1 AAU86899.1
BC034708.2 AAH34708.2
BC090932.1 AAH90932.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv564840 Chr.14:50704161 - 50733474 on Build GRCh38 Gain NIN
nsv1040775 Chr.14:50676760 - 50853094 on Build GRCh38 Gain LOC105370489 NIN

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

microtubule anchoring at centrosome
centrosome localization
centrosome-templated microtubule nucleation
calcium ion binding
protein binding
GTP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-rd95r:80/100.66.77.199:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline