This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07079213_cn
See other NDRG2 CNV Assays ›
Gene Symbol
NDRG2
Assay Reference Genome
Location

Chr.14:21023892 on build GRCh38
Cytoband
14q11.2
Assay ID Hs07079213_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57447

Gene Name:

NDRG family member 2

Gene Aliases:

SYLD

Location:

Chr.14:21016763-21070872 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NDRG2 NM_001282211.1 NP_001269140.1
NM_001282212.1 NP_001269141.1
NM_001282213.1 NP_001269142.1
NM_001282214.1 NP_001269143.1
NM_001282215.1 NP_001269144.1
NM_001282216.1 NP_001269145.1
NM_001320329.1 NP_001307258.1
NM_016250.2 NP_057334.1
NM_201535.1 NP_963293.1
NM_201536.1 NP_963294.1
NM_201537.1 NP_963831.1
NM_201538.1 NP_963832.1
NM_201539.1 NP_963833.1
NM_201540.1 NP_963834.1
NM_201541.1 NP_963835.1
XM_011536996.2 XP_011535298.1
XM_011536997.1 XP_011535299.1
XM_011536998.1 XP_011535300.1
XM_011536999.1 XP_011535301.1
XM_011537001.1 XP_011535303.1
XM_011537002.1 XP_011535304.1
XM_017021480.1 XP_016876969.1
XM_017021481.1 XP_016876970.1
XM_017021482.1 XP_016876971.1
XM_017021483.1 XP_016876972.1
XM_017021484.1 XP_016876973.1
XM_017021485.1 XP_016876974.1
XM_017021486.1 XP_016876975.1
XM_017021487.1 XP_016876976.1
XM_017021488.1 XP_016876977.1
XM_017021489.1 XP_016876978.1
XM_017021490.1 XP_016876979.1
XM_017021491.1 XP_016876980.1
XM_017021492.1 XP_016876981.1
XM_017021493.1 XP_016876982.1
XM_017021494.1 XP_016876983.1
XM_017021495.1 XP_016876984.1
XM_017021496.1 XP_016876985.1
AB033074.1
AF087872.1 AAM10500.1
AF159092.3 AAD43131.2
AF304051.1 AAL08624.1
AK024521.1
AK057843.1
AK057847.1
AK090657.1
AK096999.1
AK130029.1
AK293514.1
AW163815.1
BC010458.1 AAH10458.1
BC011240.1 AAH11240.1
BC093038.1 AAH93038.1
BG818977.1
BI759006.1
BM810623.1
BX247987.1 CAD62321.1
BX647748.1
CB153966.1
CR749252.1 CAH18108.1
DA065447.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv428293 Chr.14:20747949 - 21063624 on Build GRCh38 Gain+Loss RNASE6 RNASE2 TPPP2 RNASE13 ECRP ARHGEF40 EDDM3A MIR6717 RNASE7 RNASE3 RNASE8 NDRG2 SLC39A2 METTL17 LOC101929718 EDDM3B RNASE1
nsv832740 Chr.14:20863901 - 21076893 on Build GRCh38 Gain RNASE2 TPPP2 RNASE13 ECRP ARHGEF40 MIR6717 RNASE7 RNASE3 RNASE8 NDRG2 SLC39A2 METTL17 LOC101929718

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline