This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07105177_cn
See other GSKIP CNV Assays ›
Gene Symbol
GSKIP
Assay Reference Genome
Location

Chr.14:96372040 on build GRCh38
Cytoband
14q32.2
Assay ID Hs07105177_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51527

Gene Name:

GSK3B interacting protein

Gene Aliases:

C14orf129, HSPC210

Location:

Chr.14:96363452-96387290 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GSKIP NM_001271904.1 NP_001258833.1
NM_001271905.1 NP_001258834.1
NM_001271906.1 NP_001258835.1
NM_016472.4 NP_057556.2
XM_017021365.1 XP_016876854.1
AF151044.1 AAF36130.1
AK000796.1 BAA91380.1
AK094654.1
AK314423.1
DA488110.1
HY019704.1
HY051680.1
HY108584.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-rd95r:80/100.66.77.199:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline