This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07121617_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54046740 on build GRCh38
Cytoband
19q13.42
Assay ID Hs07121617_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
XM_017026666.1 XP_016882155.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
BQ446114.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain MIR935 CACNG8 TARM1 VSTM1 CACNG6
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion NDUFA3 OSCAR PRPF31 TARM1 TFPT VSTM1
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain MIR935 CACNG8 CACNG7 VSTM1 CACNG6
nsv1057516 Chr.19:53986233 - 54046901 on Build GRCh38 Gain CACNG8 VSTM1 CACNG6
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion ZNF761 NDUFA3 MIR372 MIR519B ZNF818P BIRC8 ZNF765 TARM1 MIR521-2 VSTM1 DPRX MIR1283-1 MIR371B LOC105372457 MIR935 RPS9 MYADM MIR519E MIR520G MIR525 FAM90A27P ZNF845 MIR515-2 MIR520H MIR512-1 MIR516B1 MIR520C MIR519A1 CACNG8 NLRP12 PRKCG TFPT MIR523 MBOAT7 MIR517C ZNF525 MIR518F MIR520F MIR521-1 MIR522 MIR517B MIR527 MIR4752 VN1R2 MIR520E MIR518D CACNG6 TMC4 LILRA5 MIR373 LILRB3 MIR526A2 MIR518B MIR516A2 MIR512-2 MIR520B MIR519D MIR516B2 MIR519A2 TSEN34 MIR371A ZNF677 MIR498 CNOT3 CACNG7 MIR520A TPM3P9 MIR520D LILRA6 MIR518C ZNF665 MIR517A MIR524 LENG1 MIR516A1 LILRB5 MIR518A1 MIR1283-2 MIR519C PRPF31 VN1R4 LOC284379 MIR518E MIR526B OSCAR LILRB2 MIR518A2 ZNF813 MIR515-1 MIR1323 ZNF331 MIR526A1
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss NDUFA3 OSCAR MIR935 CACNG8 PRPF31 TARM1 TFPT VSTM1 CACNG6

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline