This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07131846_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54049808 on build GRCh38
Cytoband
19q13.42
Assay ID Hs07131846_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
XM_017026666.1 XP_016882155.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
BQ446114.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain VSTM1 CACNG6 TARM1 MIR935 CACNG8
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion NDUFA3 VSTM1 TARM1 PRPF31 OSCAR TFPT
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain CACNG7 VSTM1 CACNG6 MIR935 CACNG8
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion ZNF818P CACNG7 MIR518F LILRA6 ZNF677 MIR517B LILRB5 MIR518C TSEN34 CNOT3 MIR515-2 MIR1323 MIR516B2 MIR526B TMC4 MIR520G MIR519A1 MIR519D OSCAR LOC105372457 MIR516A1 MIR522 MIR518A2 MYADM MIR526A2 MIR520F LILRB2 VN1R2 BIRC8 MIR518D MIR935 ZNF665 PRPF31 LOC284379 TFPT MIR520E MIR519A2 MIR515-1 MIR516B1 MIR1283-1 CACNG8 MIR512-2 ZNF761 LILRB3 ZNF845 MIR526A1 NDUFA3 PRKCG MIR373 CACNG6 TARM1 MIR498 MIR518B ZNF331 MIR520A MIR519C MIR524 MIR518A1 MIR521-1 MIR371A MIR525 MIR516A2 RPS9 VN1R4 TPM3P9 DPRX MIR512-1 MIR520D MIR520H LENG1 MIR527 MIR521-2 MIR1283-2 MIR371B VSTM1 MIR518E MBOAT7 MIR523 ZNF813 MIR520B MIR517A MIR517C MIR4752 MIR519E MIR519B MIR372 NLRP12 LILRA5 MIR520C ZNF525 FAM90A27P ZNF765
dgv6541n54 Chr.19:54049610 - 54057172 on Build GRCh38 Loss VSTM1
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss NDUFA3 VSTM1 CACNG6 TARM1 MIR935 PRPF31 OSCAR CACNG8 TFPT

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline