This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07217499_cn
See other RBCK1 CNV Assays ›
Gene Symbol
RBCK1
Assay Reference Genome
Location

Chr.20:414540 on build GRCh38
Cytoband
20p13
Assay ID Hs07217499_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10616

Gene Name:

RANBP2-type and C3HC4-type zinc finger containing 1

Gene Aliases:

C20orf18, HOIL-1, HOIL1, PBMEI, PGBM1, RBCK2, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4

Location:

Chr.20:408050-432139 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RBCK1 NM_001323956.1 NP_001310885.1
NM_001323958.1 NP_001310887.1
NM_006462.5 NP_006453.1
NM_031229.3 NP_112506.2
NR_136659.1
XM_005260645.1 XP_005260702.1
XM_011529137.1 XP_011527439.1
XM_011529138.1 XP_011527440.1
XM_011529139.2 XP_011527441.1
XM_011529140.1 XP_011527442.1
XM_017027594.1 XP_016883083.1
XM_017027595.1 XP_016883084.1
AB107766.1 BAC75409.1
AK297524.1
AK303357.1
BC015219.2 AAH15219.2
U67322.1 AAD00162.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3644953 Chr.20:397290 - 502310 on Build GRCh38 Gain CSNK2A1 TRIB3 RBCK1 TBC1D20
esv3644952 Chr.20:386013 - 1119183 on Build GRCh38 Gain ANGPT4 TRIB3 SLC52A3 PSMF1 RBCK1 LOC105372493 RSPO4 CSNK2A1 TCF15 SCRT2 SRXN1 FAM110A TBC1D20
nsv1056266 Chr.20:401152 - 522755 on Build GRCh38 Gain CSNK2A1 RBCK1 TBC1D20
esv3644956 Chr.20:405365 - 446917 on Build GRCh38 Gain RBCK1 TBC1D20
nsv510777 Chr.20:366849 - 416704 on Build GRCh38 Deletion TRIB3 RBCK1
nsv458841 Chr.20:380145 - 422066 on Build GRCh38 Loss TRIB3 RBCK1
dgv4219n100 Chr.20:80652 - 461103 on Build GRCh38 Gain DEFB128 ZCCHC3 LOC101929937 C20orf96 TRIB3 SOX12 NRSN2-AS1 DEFB127 RBCK1 DEFB129 DEFB126 NRSN2 DEFB132 TBC1D20 DEFB125
esv3893263 Chr.20:216521 - 434561 on Build GRCh38 Loss DEFB129 ZCCHC3 LOC101929937 NRSN2 C20orf96 TRIB3 DEFB132 SOX12 NRSN2-AS1 RBCK1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
I-kappaB kinase/NF-kappaB signaling
regulation of tumor necrosis factor-mediated signaling pathway
viral process
negative regulation of NF-kappaB transcription factor activity
positive regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
negative regulation of necroptotic process
protein linear polyubiquitination
positive regulation of extrinsic apoptotic signaling pathway
double-stranded DNA binding
ubiquitin-protein transferase activity
protein kinase C binding
protein binding
zinc ion binding
ligase activity
ubiquitin binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline