This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07476658_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1259955 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07476658_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 SCNN1D MIR6726 UBE2J2
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication ACAP3 TTLL10 CPSF3L MIR429 RNF223 UBE2J2 LOC105378948 MIR6726 AGRN LINC01342 LOC100288175 TAS1R3 MIR6727 TNFRSF4 TTLL10-AS1 CPTP SCNN1D TNFRSF18 C1orf159 B3GALT6 MIR200B MIR200A PUSL1 FAM132A SDF4
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss KLHL17 ACAP3 ATAD3C CPSF3L MXRA8 SLC35E2 ATAD3B MIR429 TMEM88B MRPL20 RNF223 UBE2J2 SAMD11 SLC35E2B LINC01128 MIR6726 LOC100288175 DVL1 LINC00115 MIR6727 LOC100130417 CDK11B TNFRSF4 NADK TMEM240 CPTP FAM87B SCNN1D TNFRSF18 C1orf159 HES4 MIR200B SDF4 MIR6808 ISG15 PLEKHN1 SSU72 TTLL10 FAM41C AURKAIP1 PERM1 VWA1 NOC2L LOC100287934 LOC105378948 LOC148413 AGRN ATAD3A LOC284600 LOC100288069 LOC102724312 LINC01342 TAS1R3 MMP23A ANKRD65 TTLL10-AS1 LOC107985728 FNDC10 B3GALT6 CCNL2 CDK11A MIB2 MIR200A PUSL1 FAM132A LOC107985729 MMP23B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ACAP3 CPSF3L SLC35E2 ATAD3B MIR429 TMEM88B UBE2J2 SAMD11 SLC35E2B LINC01128 MIR1302-2 OR4F29 TMEM52 LOC100132287 MIR6723 TNFRSF4 NADK TMEM240 SCNN1D TNFRSF18 C1orf159 LOC101928626 HES4 MIR200B LOC105378949 SDF4 MIR6808 LOC100506504 GABRD FAAP20 TTLL10 FAM41C LOC100133331 PERM1 MIR6859-2 LOC100287934 LOC148413 MTND2P28 AGRN LOC284600 LOC100288069 LOC102724312 TAS1R3 LOC105378592 MMP23A MIR6859-1 LOC105378947 LOC107984841 TTLL10-AS1 FNDC10 B3GALT6 MIR200A PUSL1 MTND1P23 MMP23B KLHL17 ATAD3C MXRA8 WASH7P LOC100129534 MRPL20 RNF223 MIR6726 FAM138A CALML6 LOC100288175 DVL1 LINC00115 MIR6727 PRKCZ LOC100130417 CDK11B CPTP FAM87B ISG15 PLEKHN1 SSU72 OR4F5 AURKAIP1 VWA1 NOC2L LOC729737 GNB1 LOC105378948 ATAD3A LOC100134822 LINC01342 LOC102725121 LOC105378591 OR4F16 SKI MORN1 CFAP74 ANKRD65 LOC105378589 LOC107985728 CCNL2 CDK11A MIB2 FAM132A LOC107985729 DDX11L1
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss ACAP3 SCNN1D B3GALT6 MIR6726 FAM132A SDF4 UBE2J2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss KLHL17 TTLL10 FAM41C PERM1 MIR429 NOC2L RNF223 UBE2J2 SAMD11 LOC105378948 LINC01128 AGRN LOC284600 LINC01342 LOC100288175 LINC00115 LOC100130417 TNFRSF4 TTLL10-AS1 LOC107985728 FAM87B TNFRSF18 C1orf159 HES4 B3GALT6 MIR200B MIR200A FAM132A SDF4 ISG15 PLEKHN1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion SSU72 ACAP3 ATAD3C TTLL10 CPSF3L MXRA8 AURKAIP1 ATAD3B MIR429 TMEM88B VWA1 MRPL20 UBE2J2 LOC148413 MIR6726 ATAD3A LOC102724312 LINC01342 DVL1 TAS1R3 MIR6727 CDK11B ANKRD65 TNFRSF4 TMEM240 TTLL10-AS1 CPTP SCNN1D TNFRSF18 C1orf159 FNDC10 B3GALT6 CCNL2 MIR200B MIB2 MIR200A PUSL1 FAM132A LOC107985729 SDF4 MIR6808 MMP23B
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain KLHL17 ACAP3 TTLL10 CPSF3L MXRA8 AURKAIP1 PERM1 MIR429 TMEM88B NOC2L MRPL20 RNF223 UBE2J2 SAMD11 LOC105378948 LOC148413 MIR6726 AGRN LOC284600 LINC01342 LOC100288175 DVL1 TAS1R3 MIR6727 LOC100130417 ANKRD65 TNFRSF4 TTLL10-AS1 CPTP LOC107985728 SCNN1D TNFRSF18 C1orf159 HES4 B3GALT6 CCNL2 MIR200B MIR200A PUSL1 FAM132A SDF4 MIR6808 ISG15 PLEKHN1
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10 MIR429 TNFRSF4 RNF223 TTLL10-AS1 UBE2J2 LOC105378948 SCNN1D TNFRSF18 C1orf159 B3GALT6 MIR200B MIR200A FAM132A SDF4 LINC01342
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss ACAP3 TTLL10 MIR429 TNFRSF4 RNF223 TTLL10-AS1 UBE2J2 SCNN1D TNFRSF18 C1orf159 B3GALT6 MIR6726 MIR200B MIR200A FAM132A SDF4 LINC01342
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain ACAP3 TTLL10 CPSF3L MIR6727 MIR429 TNFRSF4 TTLL10-AS1 UBE2J2 SCNN1D TNFRSF18 B3GALT6 MIR6726 MIR200B MIR200A PUSL1 FAM132A SDF4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline