This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07476875_cn
See other CPTP CNV Assays ›
Gene Symbol
CPTP
Assay Reference Genome
Location

Chr.1:1325176 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07476875_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

80772

Gene Name:

ceramide-1-phosphate transfer protein

Gene Aliases:

GLTPD1

Location:

Chr.1:1324756-1328896 on Build GRCh38

Assay Gene Location:

Overlaps Intron 1 - Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPTP NM_001029885.1 NP_001025056.1
XM_005244801.3 XP_005244858.1
XM_005244802.1 XP_005244859.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss NOC2L LOC148413 OR4F16 LOC107985728 TTLL10 LOC100288069 MIR200B DVL1 LOC729737 MIR6808 UBE2J2 TMEM240 SSU72 FAM87B CDK11B ANKRD65 DDX11L1 MIR1302-2 MIR429 LOC107985729 LOC105378948 MIR200A LOC105378589 LOC100129534 HES4 MIR6727 MTND1P23 LOC102725121 SCNN1D LOC100506504 SDF4 KLHL17 C1orf159 MTND2P28 LOC100130417 NADK FAM132A MIR6859-2 TMEM52 MIB2 WASH7P FAAP20 TNFRSF4 LOC105378591 LINC01128 LOC100288175 LOC100133331 SLC35E2B CPTP MIR6723 LOC105378949 PUSL1 MXRA8 LOC105378592 LOC100134822 CCNL2 LINC01342 TMEM88B B3GALT6 LINC00115 MORN1 ATAD3B LOC284600 AGRN FAM41C CPSF3L TAS1R3 ACAP3 MMP23A FNDC10 PERM1 TTLL10-AS1 CDK11A VWA1 ATAD3C SLC35E2 GABRD LOC101928626 LOC105378947 ATAD3A CALML6 PRKCZ PLEKHN1 MMP23B LOC102724312 GNB1 AURKAIP1 TNFRSF18 LOC100132287 MIR6726 OR4F5 MIR6859-1 LOC107984841 FAM138A ISG15 MRPL20 SKI LOC100287934 CFAP74 RNF223 SAMD11 OR4F29
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 MIR6727 MIR6726
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 AURKAIP1 MIR6727 MXRA8 MIR6726 SCNN1D
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 AURKAIP1 MIR6727 MXRA8
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss CPTP TTLL10 MIR200B DVL1 PUSL1 MIR6727 MXRA8 LINC01342 SCNN1D MIR6808 B3GALT6 UBE2J2 SDF4 C1orf159 AURKAIP1 TNFRSF18 MIR6726 CPSF3L FAM132A TAS1R3 ACAP3 TTLL10-AS1 MIR429 RNF223 TNFRSF4 MIR200A
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 MIR6727 MXRA8
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPSF3L CPTP
nsv544972 Chr.1:1321172 - 1337736 on Build GRCh38 Loss CPSF3L CPTP TAS1R3 DVL1
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 AURKAIP1 MIR6727 MXRA8 MIR6726 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication CPTP TTLL10 MIR200B PUSL1 MIR6727 LINC01342 SCNN1D B3GALT6 UBE2J2 SDF4 C1orf159 AGRN TNFRSF18 MIR6726 CPSF3L FAM132A TAS1R3 ACAP3 TTLL10-AS1 MIR429 RNF223 TNFRSF4 LOC105378948 LOC100288175 MIR200A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss SLC35E2B NOC2L CPTP LOC148413 LOC107985728 TTLL10 LOC100288069 MIR200B DVL1 PUSL1 MXRA8 CCNL2 LINC01342 MIR6808 TMEM88B B3GALT6 LINC00115 UBE2J2 TMEM240 ATAD3B LOC284600 SSU72 AGRN FAM87B FAM41C CDK11B CPSF3L ANKRD65 TAS1R3 ACAP3 MMP23A FNDC10 PERM1 TTLL10-AS1 MIR429 LOC107985729 CDK11A VWA1 LOC105378948 MIR200A ATAD3C SLC35E2 HES4 ATAD3A MIR6727 SCNN1D PLEKHN1 SDF4 KLHL17 MMP23B LOC102724312 C1orf159 LOC100130417 AURKAIP1 TNFRSF18 MIR6726 NADK FAM132A MIB2 ISG15 MRPL20 LOC100287934 RNF223 TNFRSF4 LINC01128 SAMD11 LOC100288175
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 MIR6727 MXRA8
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6808 CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 MIR6727 MXRA8 MIR6726 SCNN1D
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion CPTP LOC148413 TTLL10 MIR200B DVL1 PUSL1 ATAD3A MIR6727 MXRA8 CCNL2 LINC01342 SCNN1D MIR6808 TMEM88B B3GALT6 UBE2J2 SDF4 TMEM240 MMP23B LOC102724312 ATAD3B C1orf159 SSU72 AURKAIP1 TNFRSF18 CDK11B MIR6726 CPSF3L ANKRD65 FAM132A TAS1R3 ACAP3 MIB2 FNDC10 TTLL10-AS1 MRPL20 MIR429 LOC107985729 TNFRSF4 VWA1 MIR200A ATAD3C
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss CPSF3L CPTP TAS1R3 ACAP3 DVL1 PUSL1 MIR6727 MXRA8 SCNN1D MIR6808 UBE2J2 AURKAIP1 MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain NOC2L CPTP LOC148413 LOC107985728 TTLL10 MIR200B HES4 DVL1 PUSL1 MIR6727 MXRA8 CCNL2 LINC01342 SCNN1D MIR6808 TMEM88B B3GALT6 PLEKHN1 UBE2J2 SDF4 KLHL17 LOC284600 C1orf159 LOC100130417 AGRN AURKAIP1 TNFRSF18 MIR6726 CPSF3L ANKRD65 FAM132A TAS1R3 ACAP3 PERM1 TTLL10-AS1 ISG15 MRPL20 MIR429 RNF223 TNFRSF4 LOC105378948 SAMD11 LOC100288175 MIR200A

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0