This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07476983_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1260059 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07476983_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 SCNN1D MIR6726 UBE2J2
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication LOC100288175 MIR200B B3GALT6 MIR6726 TNFRSF18 RNF223 TTLL10 MIR6727 MIR429 C1orf159 CPTP CPSF3L TTLL10-AS1 LINC01342 PUSL1 MIR200A FAM132A SDF4 UBE2J2 ACAP3 TAS1R3 SCNN1D TNFRSF4 AGRN LOC105378948
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss SLC35E2B LOC100288175 AURKAIP1 SAMD11 DVL1 HES4 TNFRSF18 LINC00115 TTLL10 FAM87B FNDC10 MXRA8 MIR6727 LOC148413 CDK11B C1orf159 TMEM88B CPSF3L MRPL20 LOC102724312 TTLL10-AS1 KLHL17 MIR200A FAM132A SDF4 UBE2J2 ATAD3B SCNN1D VWA1 LOC100130417 MIR200B CDK11A LOC107985728 MIB2 ISG15 B3GALT6 MIR6726 TMEM240 NOC2L RNF223 LOC107985729 MMP23B ATAD3A PERM1 ANKRD65 ATAD3C LOC284600 MIR429 PLEKHN1 CPTP LINC01128 NADK FAM41C LINC01342 PUSL1 MMP23A LOC100287934 MIR6808 ACAP3 CCNL2 SLC35E2 SSU72 TAS1R3 TNFRSF4 AGRN LOC105378948 LOC100288069
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SLC35E2B LOC100288175 AURKAIP1 TNFRSF18 LOC105378589 TTLL10 FAM87B FNDC10 OR4F5 MXRA8 LOC729737 CDK11B C1orf159 LOC105378949 LOC102724312 LOC105378592 MORN1 MIR200A ATAD3B PRKCZ LOC105378947 LOC101928626 LOC100130417 MIR200B CDK11A FAM138A ISG15 B3GALT6 CALML6 MIR6726 LOC107984841 LOC105378591 ATAD3A PERM1 ATAD3C MIR429 NADK FAM41C MIR6859-2 LOC100287934 MIR6808 GABRD MTND2P28 SKI DDX11L1 OR4F29 OR4F16 LOC100133331 SAMD11 DVL1 HES4 MIR6723 LINC00115 FAAP20 MIR6727 LOC148413 LOC102725121 TMEM88B CPSF3L MRPL20 TTLL10-AS1 LOC100129534 KLHL17 FAM132A SDF4 UBE2J2 LOC100132287 SCNN1D MTND1P23 VWA1 TMEM52 MIR1302-2 LOC107985728 MIB2 TMEM240 NOC2L RNF223 LOC107985729 MMP23B ANKRD65 LOC284600 PLEKHN1 CPTP LINC01128 LOC100506504 LOC100134822 LINC01342 PUSL1 MMP23A ACAP3 CCNL2 SLC35E2 SSU72 CFAP74 TAS1R3 TNFRSF4 MIR6859-1 GNB1 WASH7P AGRN LOC105378948 LOC100288069
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss ACAP3 B3GALT6 SCNN1D MIR6726 FAM132A SDF4 UBE2J2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC100288175 MIR200B LOC107985728 ISG15 B3GALT6 SAMD11 HES4 TNFRSF18 NOC2L RNF223 LINC00115 TTLL10 FAM87B PERM1 LOC284600 MIR429 C1orf159 PLEKHN1 LINC01128 TTLL10-AS1 FAM41C LINC01342 KLHL17 MIR200A FAM132A SDF4 UBE2J2 TNFRSF4 AGRN LOC100130417 LOC105378948
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion AURKAIP1 MIR200B MIB2 B3GALT6 DVL1 MIR6726 TNFRSF18 TMEM240 LOC107985729 TTLL10 FNDC10 MMP23B ATAD3A MXRA8 ANKRD65 ATAD3C MIR6727 MIR429 LOC148413 CDK11B C1orf159 CPTP TMEM88B CPSF3L MRPL20 LOC102724312 TTLL10-AS1 LINC01342 PUSL1 MIR200A FAM132A MIR6808 SDF4 UBE2J2 ACAP3 CCNL2 ATAD3B SSU72 TAS1R3 SCNN1D TNFRSF4 VWA1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC100288175 AURKAIP1 MIR200B LOC107985728 ISG15 B3GALT6 SAMD11 DVL1 HES4 MIR6726 TNFRSF18 NOC2L RNF223 TTLL10 PERM1 MXRA8 ANKRD65 MIR6727 LOC284600 MIR429 LOC148413 C1orf159 PLEKHN1 CPTP TMEM88B CPSF3L MRPL20 TTLL10-AS1 LINC01342 PUSL1 KLHL17 MIR200A FAM132A MIR6808 SDF4 UBE2J2 ACAP3 CCNL2 TAS1R3 SCNN1D TNFRSF4 AGRN LOC100130417 LOC105378948
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR200B B3GALT6 TTLL10-AS1 TNFRSF18 LINC01342 MIR200A FAM132A RNF223 SDF4 UBE2J2 TTLL10 SCNN1D TNFRSF4 MIR429 C1orf159 LOC105378948
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR200B B3GALT6 TTLL10-AS1 MIR6726 TNFRSF18 LINC01342 MIR200A FAM132A RNF223 SDF4 UBE2J2 ACAP3 TTLL10 SCNN1D TNFRSF4 MIR429 C1orf159
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L MIR200B B3GALT6 TTLL10-AS1 MIR6726 TNFRSF18 PUSL1 MIR200A FAM132A SDF4 UBE2J2 ACAP3 TTLL10 SCNN1D TNFRSF4 MIR6727 MIR429

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline