This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07476994_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1260135 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07476994_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 UBE2J2 MIR6726 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication PUSL1 UBE2J2 TAS1R3 LOC100288175 MIR6727 MIR429 LOC105378948 TTLL10 FAM132A MIR6726 SCNN1D SDF4 MIR200A TNFRSF18 CPSF3L TNFRSF4 MIR200B CPTP C1orf159 ACAP3 LINC01342 B3GALT6 AGRN RNF223 TTLL10-AS1
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss DVL1 ATAD3C CDK11A LOC100288069 MXRA8 ATAD3B TTLL10 MIB2 SDF4 TMEM240 MIR200A TNFRSF18 CPSF3L TNFRSF4 MIR200B CCNL2 MIR6808 VWA1 MRPL20 TMEM88B LOC102724312 ISG15 LINC00115 MMP23A LOC148413 CDK11B FAM87B B3GALT6 SSU72 ATAD3A RNF223 TTLL10-AS1 ANKRD65 PUSL1 UBE2J2 SLC35E2 TAS1R3 LOC100287934 LOC100288175 MIR6727 MIR429 LOC105378948 SLC35E2B HES4 SAMD11 NOC2L FAM132A FAM41C FNDC10 MIR6726 SCNN1D PERM1 LOC107985729 CPTP KLHL17 NADK LOC107985728 LOC284600 C1orf159 LINC01128 MMP23B AURKAIP1 ACAP3 PLEKHN1 LOC100130417 LINC01342 AGRN
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ATAD3C LOC105378589 CDK11A ATAD3B DDX11L1 MIR200A TNFRSF18 CPSF3L MIR6808 TMEM88B LOC102724312 MIR6723 MMP23A LOC148413 GABRD FAM87B SSU72 ATAD3A SLC35E2 LOC100506504 MIR429 SLC35E2B FAM132A FAM41C FAAP20 MIR6726 OR4F5 LOC105378592 OR4F16 CPTP NADK LOC105378591 LOC107985728 CFAP74 LOC284600 MTND2P28 MMP23B WASH7P MIR6859-1 SKI ACAP3 FAM138A LOC107984841 DVL1 LOC100288069 MXRA8 TTLL10 OR4F29 MIB2 SDF4 TMEM240 PRKCZ LOC100133331 TNFRSF4 MIR200B LOC105378947 CCNL2 VWA1 MRPL20 ISG15 LINC00115 CDK11B LOC100134822 B3GALT6 GNB1 RNF223 TTLL10-AS1 ANKRD65 PUSL1 UBE2J2 CALML6 LOC101928626 TAS1R3 LOC100287934 LOC100288175 TMEM52 MIR1302-2 MIR6727 LOC105378948 HES4 SAMD11 NOC2L FNDC10 MIR6859-2 SCNN1D PERM1 LOC107985729 KLHL17 LOC102725121 MTND1P23 LOC100129534 LOC105378949 C1orf159 LINC01128 LOC729737 AURKAIP1 MORN1 LOC100132287 PLEKHN1 LOC100130417 LINC01342 AGRN
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss ACAP3 UBE2J2 FAM132A MIR6726 SCNN1D SDF4 B3GALT6
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss UBE2J2 LOC100288175 MIR429 LOC105378948 TTLL10 HES4 SAMD11 NOC2L FAM132A FAM41C SDF4 MIR200A PERM1 TNFRSF18 TNFRSF4 MIR200B KLHL17 LOC107985728 LOC284600 C1orf159 LINC01128 ISG15 LINC00115 FAM87B PLEKHN1 LOC100130417 LINC01342 B3GALT6 AGRN RNF223 TTLL10-AS1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ANKRD65 PUSL1 UBE2J2 TAS1R3 DVL1 ATAD3C MIR6727 MIR429 MXRA8 ATAD3B TTLL10 FAM132A MIB2 FNDC10 MIR6726 SCNN1D SDF4 TMEM240 MIR200A TNFRSF18 CPSF3L TNFRSF4 LOC107985729 MIR200B CPTP CCNL2 MIR6808 C1orf159 VWA1 MRPL20 TMEM88B LOC102724312 MMP23B LOC148413 AURKAIP1 ACAP3 CDK11B LINC01342 B3GALT6 SSU72 ATAD3A TTLL10-AS1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain ANKRD65 PUSL1 UBE2J2 TAS1R3 LOC100288175 DVL1 MIR6727 MIR429 MXRA8 LOC105378948 TTLL10 HES4 SAMD11 NOC2L FAM132A MIR6726 SCNN1D SDF4 MIR200A PERM1 TNFRSF18 CPSF3L TNFRSF4 MIR200B CPTP KLHL17 CCNL2 LOC107985728 MIR6808 LOC284600 C1orf159 MRPL20 TMEM88B ISG15 LOC148413 AURKAIP1 ACAP3 PLEKHN1 LOC100130417 LINC01342 B3GALT6 AGRN RNF223 TTLL10-AS1
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain UBE2J2 C1orf159 MIR429 LOC105378948 TTLL10 FAM132A SCNN1D SDF4 MIR200A LINC01342 B3GALT6 TNFRSF18 TNFRSF4 RNF223 MIR200B TTLL10-AS1
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss UBE2J2 C1orf159 MIR429 ACAP3 TTLL10 FAM132A MIR6726 SCNN1D SDF4 MIR200A LINC01342 B3GALT6 TNFRSF18 TNFRSF4 RNF223 MIR200B TTLL10-AS1
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain PUSL1 UBE2J2 MIR6727 MIR429 ACAP3 TTLL10 FAM132A MIR6726 SCNN1D SDF4 MIR200A B3GALT6 TNFRSF18 CPSF3L TNFRSF4 MIR200B TTLL10-AS1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline