This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07477006_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1259457 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07477006_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 ACAP3 SCNN1D MIR6726
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication CPTP MIR429 LOC105378948 TTLL10-AS1 FAM132A LOC100288175 UBE2J2 TNFRSF4 SCNN1D LINC01342 AGRN CPSF3L C1orf159 RNF223 SDF4 MIR200B MIR6727 MIR200A TNFRSF18 ACAP3 TAS1R3 B3GALT6 TTLL10 PUSL1 MIR6726
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC102724312 CPTP LOC284600 LOC105378948 LOC100287934 TMEM88B TTLL10-AS1 DVL1 FAM132A HES4 MRPL20 UBE2J2 TNFRSF4 LINC01128 LOC148413 AGRN ATAD3B CCNL2 LOC107985728 CPSF3L SLC35E2B NOC2L C1orf159 NADK SDF4 MIR200B VWA1 MIR6727 MMP23A ISG15 AURKAIP1 ANKRD65 TNFRSF18 ACAP3 MIR6808 TAS1R3 B3GALT6 MIR6726 ATAD3C MIR429 PLEKHN1 LOC100288175 SLC35E2 MMP23B KLHL17 ATAD3A LOC100130417 SCNN1D LINC01342 TMEM240 FAM41C CDK11A SAMD11 CDK11B MIB2 LOC100288069 RNF223 FNDC10 SSU72 PERM1 LOC107985729 FAM87B MIR200A LINC00115 MXRA8 TTLL10 PUSL1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CPTP CALML6 LOC105378948 TMEM88B TTLL10-AS1 PRKCZ LOC105378949 TNFRSF4 MIR6859-1 MORN1 CCNL2 LOC107985728 CPSF3L OR4F5 CFAP74 NOC2L MTND1P23 LOC105378591 SDF4 MIR200B OR4F29 MIR6727 MMP23A AURKAIP1 LOC100134822 ACAP3 MIR6808 TAS1R3 ATAD3C FAAP20 LOC101928626 PLEKHN1 LOC100288175 GNB1 SLC35E2 MMP23B LOC100129534 LOC100130417 SCNN1D GABRD FAM41C CDK11A SAMD11 LOC102725121 MIB2 LOC100288069 RNF223 DDX11L1 SSU72 PERM1 LOC107985729 OR4F16 MIR200A LINC00115 MXRA8 PUSL1 LOC102724312 LOC284600 LOC100287934 LOC105378592 DVL1 FAM132A HES4 MRPL20 UBE2J2 LINC01128 LOC100506504 LOC105378947 LOC148413 WASH7P AGRN LOC729737 ATAD3B SLC35E2B C1orf159 NADK TMEM52 VWA1 ISG15 ANKRD65 TNFRSF18 B3GALT6 MIR1302-2 MIR6726 MIR6723 MIR429 LOC100133331 LOC107984841 MIR6859-2 KLHL17 ATAD3A LINC01342 TMEM240 CDK11B SKI FNDC10 FAM87B LOC100132287 MTND2P28 FAM138A TTLL10 LOC105378589
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 ACAP3 SCNN1D B3GALT6 MIR6726 SDF4 FAM132A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC284600 MIR429 LOC105378948 PLEKHN1 TTLL10-AS1 FAM132A LOC100288175 HES4 UBE2J2 TNFRSF4 KLHL17 LINC01128 LOC100130417 LINC01342 AGRN LOC107985728 FAM41C SAMD11 NOC2L C1orf159 RNF223 SDF4 MIR200B PERM1 FAM87B ISG15 MIR200A TNFRSF18 LINC00115 B3GALT6 TTLL10
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion LOC102724312 CPTP ATAD3C MIR429 TMEM88B TTLL10-AS1 DVL1 FAM132A MRPL20 MMP23B UBE2J2 TNFRSF4 ATAD3A LOC148413 SCNN1D LINC01342 TMEM240 ATAD3B CCNL2 CPSF3L CDK11B C1orf159 MIB2 FNDC10 SDF4 SSU72 MIR200B VWA1 MIR6727 LOC107985729 AURKAIP1 MIR200A ANKRD65 TNFRSF18 ACAP3 MIR6808 TAS1R3 B3GALT6 MXRA8 TTLL10 PUSL1 MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain CPTP LOC284600 MIR429 LOC105378948 TMEM88B PLEKHN1 TTLL10-AS1 DVL1 FAM132A LOC100288175 HES4 MRPL20 UBE2J2 TNFRSF4 KLHL17 LOC148413 LOC100130417 SCNN1D LINC01342 AGRN CCNL2 LOC107985728 CPSF3L SAMD11 NOC2L C1orf159 RNF223 SDF4 MIR200B PERM1 MIR6727 ISG15 AURKAIP1 MIR200A ANKRD65 TNFRSF18 ACAP3 MIR6808 TAS1R3 B3GALT6 MXRA8 TTLL10 PUSL1 MIR6726
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR429 LOC105378948 C1orf159 TTLL10-AS1 RNF223 SDF4 FAM132A MIR200B MIR200A UBE2J2 TNFRSF4 TNFRSF18 SCNN1D B3GALT6 LINC01342 TTLL10
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR429 C1orf159 TTLL10-AS1 RNF223 SDF4 FAM132A MIR200B MIR200A UBE2J2 TNFRSF4 TNFRSF18 ACAP3 SCNN1D B3GALT6 LINC01342 TTLL10 MIR6726
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L MIR429 TTLL10-AS1 SDF4 FAM132A MIR200B MIR6727 MIR200A UBE2J2 TNFRSF4 TNFRSF18 ACAP3 SCNN1D B3GALT6 TTLL10 PUSL1 MIR6726

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline