This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07477020_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1618098 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07477020_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK123400.1 2 1709
AK128167.1
AK128553.1
AK293642.1
AK302757.1
AK309362.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1
DA391220.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MIB2 CDK11B CDK11A GNB1 MMP23B NADK MMP23A SLC35E2B SLC35E2 FNDC10
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss MIB2 CDK11B CDK11A MMP23B SSU72 MMP23A SLC35E2B SLC35E2 FNDC10
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss UBE2J2 MIB2 DVL1 LOC148413 KLHL17 HES4 TMEM88B ATAD3B SLC35E2B TMEM240 RNF223 LOC107985728 SDF4 AURKAIP1 C1orf159 MMP23A LOC100287934 VWA1 PLEKHN1 ATAD3C SAMD11 CPTP NADK ISG15 AGRN LINC01342 LOC100130417 TNFRSF18 MIR6808 FAM41C LOC102724312 FAM132A LOC284600 SLC35E2 MXRA8 MRPL20 CCNL2 B3GALT6 CDK11B LOC100288069 PERM1 TTLL10-AS1 LINC01128 ANKRD65 MIR200A LOC100288175 TNFRSF4 MMP23B FNDC10 CPSF3L TAS1R3 MIR200B CDK11A MIR6727 NOC2L FAM87B LOC105378948 MIR6726 SSU72 ATAD3A LOC107985729 MIR429 ACAP3 PUSL1 SCNN1D TTLL10 LINC00115
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss DVL1 DDX11L1 TMEM52 LOC148413 KLHL17 GNB1 TMEM88B TMEM240 SKI MTND2P28 SDF4 MMP23A LOC100506504 FAM138A LOC100287934 MIR6859-1 NADK LOC100130417 TNFRSF18 MIR6808 FAM132A OR4F29 LOC284600 SLC35E2 FAAP20 MXRA8 LOC100129534 PERM1 TTLL10-AS1 LOC729737 MIR6859-2 MIR200A LOC100288175 TNFRSF4 MMP23B CPSF3L TAS1R3 LOC105378947 MIR200B CDK11A NOC2L MIR6723 SSU72 ATAD3A MTND1P23 MIR429 LOC105378949 PRKCZ ACAP3 SCNN1D WASH7P UBE2J2 MIB2 LOC100134822 LOC105378589 HES4 ATAD3B SLC35E2B RNF223 LOC107985728 AURKAIP1 C1orf159 OR4F16 LOC105378592 VWA1 PLEKHN1 ATAD3C SAMD11 CPTP GABRD ISG15 AGRN LINC01342 LOC105378591 FAM41C OR4F5 LOC102724312 MRPL20 CCNL2 B3GALT6 CDK11B LOC100288069 CALML6 LINC01128 LOC100132287 ANKRD65 MIR1302-2 FNDC10 LOC107984841 LOC100133331 MORN1 MIR6727 FAM87B CFAP74 LOC105378948 MIR6726 LOC101928626 LOC107985729 LOC102725121 PUSL1 TTLL10 LINC00115
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MIB2 CDK11B CDK11A GNB1 MMP23B SSU72 NADK MMP23A SLC35E2B SLC35E2 FNDC10
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion UBE2J2 MIB2 DVL1 B3GALT6 LOC148413 CDK11B TMEM88B ATAD3B TTLL10-AS1 TMEM240 ANKRD65 SDF4 AURKAIP1 C1orf159 MIR200A TNFRSF4 MMP23B FNDC10 CPSF3L TAS1R3 VWA1 MIR200B ATAD3C MIR6727 CPTP MIR6726 SSU72 ATAD3A LOC107985729 LINC01342 MIR429 TNFRSF18 MIR6808 ACAP3 PUSL1 SCNN1D LOC102724312 TTLL10 FAM132A MXRA8 MRPL20 CCNL2
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss MIB2 CDK11B CDK11A MMP23B SSU72 NADK MMP23A SLC35E2B SLC35E2 FNDC10
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss MIB2 CDK11B MMP23B SLC35E2B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MIB2 TMEM52 LOC148413 CDK11B LOC105378589 GNB1 TMEM88B ATAD3B SLC35E2B TMEM240 CALML6 ANKRD65 MMP23B MMP23A FNDC10 VWA1 ATAD3C CDK11A CFAP74 GABRD SSU72 NADK ATAD3A LOC107985729 LOC105378949 LOC105378591 PRKCZ LOC102724312 SLC35E2 MRPL20 CCNL2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline