This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07477212_cn
See other CPTP CNV Assays ›
Gene Symbol
CPTP
Assay Reference Genome
Location

Chr.1:1325273 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07477212_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

80772

Gene Name:

ceramide-1-phosphate transfer protein

Gene Aliases:

GLTPD1

Location:

Chr.1:1324756-1328896 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPTP NM_001029885.1 NP_001025056.1
XM_005244801.3 1 131 XP_005244858.1
XM_005244802.1 XP_005244859.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss WASH7P LOC100130417 LINC00115 FAM41C MIR6859-2 LOC100129534 CFAP74 FAAP20 SKI LOC105378948 FAM87B TTLL10 CDK11B MIR6808 AGRN MORN1 MIR6726 LOC100287934 RNF223 NADK PERM1 PLEKHN1 OR4F16 UBE2J2 ATAD3B MIR200A TMEM88B LOC100506504 CCNL2 CPTP ATAD3A LOC105378589 MRPL20 OR4F5 LOC105378592 MIR1302-2 NOC2L PRKCZ MMP23A SDF4 MXRA8 B3GALT6 LOC107985729 SLC35E2B TMEM240 LINC01342 MIR6859-1 MIR6723 FNDC10 TTLL10-AS1 SAMD11 ATAD3C ACAP3 GNB1 LOC105378949 SLC35E2 LOC100134822 LOC100288069 TNFRSF4 TNFRSF18 MIB2 LOC100132287 FAM132A TAS1R3 LOC102724312 LOC100133331 LOC107984841 MTND1P23 SSU72 DVL1 FAM138A TMEM52 CPSF3L GABRD MIR6727 CALML6 OR4F29 CDK11A DDX11L1 MMP23B LOC107985728 HES4 SCNN1D MIR429 LOC102725121 LINC01128 PUSL1 LOC100288175 LOC148413 ISG15 LOC105378591 LOC729737 VWA1 LOC101928626 ANKRD65 KLHL17 MIR200B LOC284600 AURKAIP1 MTND2P28 C1orf159 LOC105378947
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss DVL1 TAS1R3 CPSF3L MIR6808 MIR6727 CPTP ACAP3 PUSL1 MIR6726
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss DVL1 TAS1R3 CPSF3L AURKAIP1 MIR6808 MIR6727 CPTP ACAP3 PUSL1 SCNN1D MIR6726 MXRA8
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain DVL1 TAS1R3 CPSF3L AURKAIP1 MIR6808 MIR6727 CPTP ACAP3 PUSL1 MXRA8
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss TTLL10-AS1 UBE2J2 MIR200A ACAP3 SCNN1D MIR429 CPTP PUSL1 TNFRSF4 TNFRSF18 FAM132A TTLL10 TAS1R3 MIR6808 SDF4 MIR6726 MXRA8 B3GALT6 RNF223 DVL1 MIR200B CPSF3L AURKAIP1 MIR6727 LINC01342 C1orf159
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss DVL1 TAS1R3 CPSF3L MIR6808 MIR6727 CPTP ACAP3 PUSL1 MXRA8
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPSF3L CPTP
nsv544972 Chr.1:1321172 - 1337736 on Build GRCh38 Loss DVL1 TAS1R3 CPSF3L CPTP
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion DVL1 TAS1R3 CPSF3L AURKAIP1 MIR6808 MIR6727 CPTP ACAP3 PUSL1 SCNN1D MIR6726 MXRA8
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TTLL10-AS1 UBE2J2 MIR200A ACAP3 SCNN1D MIR429 CPTP PUSL1 TNFRSF4 TNFRSF18 LOC100288175 LOC105378948 FAM132A TTLL10 TAS1R3 AGRN SDF4 MIR6726 B3GALT6 RNF223 MIR200B CPSF3L MIR6727 LINC01342 C1orf159
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss TTLL10-AS1 SAMD11 LOC100130417 ATAD3C LINC00115 FAM41C ACAP3 SLC35E2 LOC100288069 TNFRSF4 TNFRSF18 MIB2 LOC105378948 FAM87B FAM132A TTLL10 TAS1R3 CDK11B LOC102724312 MIR6808 AGRN MIR6726 LOC100287934 RNF223 SSU72 NADK PERM1 DVL1 PLEKHN1 CPSF3L MIR6727 CDK11A UBE2J2 ATAD3B MIR200A MMP23B LOC107985728 HES4 SCNN1D MIR429 TMEM88B LINC01128 CCNL2 CPTP ATAD3A PUSL1 MRPL20 LOC100288175 LOC148413 ISG15 VWA1 NOC2L MMP23A SDF4 MXRA8 B3GALT6 LOC107985729 SLC35E2B ANKRD65 TMEM240 KLHL17 MIR200B LOC284600 AURKAIP1 LINC01342 C1orf159 FNDC10
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss DVL1 TAS1R3 CPSF3L MIR6808 MIR6727 CPTP ACAP3 PUSL1 MXRA8
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain DVL1 TAS1R3 CPSF3L MIR6808 MIR6727 CPTP ACAP3 PUSL1 SCNN1D MIR6726 MXRA8
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion TTLL10-AS1 UBE2J2 ATAD3B ATAD3C MIR200A MMP23B ACAP3 SCNN1D MIR429 TMEM88B CCNL2 CPTP ATAD3A PUSL1 TNFRSF4 MRPL20 TNFRSF18 MIB2 LOC148413 FAM132A TTLL10 TAS1R3 CDK11B LOC102724312 VWA1 MIR6808 SDF4 MIR6726 MXRA8 B3GALT6 LOC107985729 ANKRD65 SSU72 TMEM240 DVL1 MIR200B CPSF3L AURKAIP1 MIR6727 LINC01342 C1orf159 FNDC10
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss TAS1R3 UBE2J2 MIR6808 ACAP3 SCNN1D MIR6726 MXRA8 DVL1 CPSF3L AURKAIP1 MIR6727 CPTP PUSL1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TTLL10-AS1 UBE2J2 SAMD11 LOC100130417 MIR200A ACAP3 LOC107985728 HES4 SCNN1D MIR429 TMEM88B CCNL2 CPTP PUSL1 TNFRSF4 MRPL20 TNFRSF18 LOC100288175 LOC105378948 LOC148413 FAM132A TTLL10 TAS1R3 ISG15 NOC2L MIR6808 AGRN SDF4 MIR6726 MXRA8 B3GALT6 RNF223 ANKRD65 PERM1 KLHL17 DVL1 PLEKHN1 MIR200B CPSF3L LOC284600 AURKAIP1 MIR6727 LINC01342 C1orf159

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ln7zh:80/100.66.79.20:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0