This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07477574_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1261521 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07477574_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 SCNN1D MIR6726 ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR200B LINC01342 TTLL10 TAS1R3 RNF223 MIR429 LOC100288175 UBE2J2 B3GALT6 CPSF3L ACAP3 SDF4 LOC105378948 TNFRSF18 MIR6726 C1orf159 TTLL10-AS1 MIR200A MIR6727 SCNN1D FAM132A CPTP AGRN TNFRSF4 PUSL1
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MIR200B ISG15 TTLL10 LOC100288175 TMEM88B FNDC10 LOC100288069 UBE2J2 ATAD3B FAM41C ATAD3C SSU72 B3GALT6 CPSF3L LOC100130417 MXRA8 TMEM240 LOC102724312 PERM1 ANKRD65 TTLL10-AS1 MIR6727 KLHL17 LINC00115 VWA1 SLC35E2B LOC284600 LOC100287934 LINC01342 FAM87B TAS1R3 SLC35E2 RNF223 MIR429 NADK LOC107985729 HES4 CDK11A CDK11B CCNL2 SAMD11 AURKAIP1 ACAP3 LOC107985728 SDF4 LOC105378948 MIR6808 TNFRSF18 MIR6726 MMP23B MMP23A LOC148413 C1orf159 DVL1 MIR200A SCNN1D NOC2L FAM132A ATAD3A LINC01128 CPTP AGRN TNFRSF4 PLEKHN1 MRPL20 MIB2 PUSL1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ISG15 TTLL10 LOC100288175 MTND1P23 SKI FNDC10 LOC100288069 UBE2J2 SSU72 B3GALT6 LOC100133331 MIR6859-1 CPSF3L MXRA8 TMEM240 LOC102724312 OR4F16 ANKRD65 MIR6727 GABRD KLHL17 LINC00115 VWA1 SLC35E2B LOC100287934 LOC100134822 TAS1R3 LOC105378591 SLC35E2 RNF223 MIR429 LOC100132287 NADK LOC105378592 CALML6 HES4 CDK11B CCNL2 ACAP3 LOC105378948 MIR6808 TNFRSF18 MIR6726 MMP23A C1orf159 SCNN1D ATAD3A CPTP AGRN MIB2 PUSL1 MIR200B FAAP20 FAM138A TMEM88B LOC102725121 OR4F5 ATAD3B OR4F29 FAM41C ATAD3C LOC100130417 MIR1302-2 PRKCZ CFAP74 PERM1 LOC105378589 TTLL10-AS1 MIR6723 LOC100129534 LOC284600 LINC01342 FAM87B LOC729737 MTND2P28 LOC107985729 TMEM52 CDK11A MORN1 SAMD11 LOC101928626 DDX11L1 AURKAIP1 MIR6859-2 LOC107985728 SDF4 LOC105378949 GNB1 MMP23B LOC148413 LOC107984841 DVL1 WASH7P MIR200A LOC100506504 NOC2L FAM132A LINC01128 LOC105378947 TNFRSF4 PLEKHN1 MRPL20
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 SCNN1D FAM132A B3GALT6 MIR6726 ACAP3 SDF4
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC284600 MIR200B ISG15 LINC01342 TTLL10 FAM87B RNF223 MIR429 LOC100288175 UBE2J2 FAM41C HES4 B3GALT6 SAMD11 LOC100130417 LOC107985728 SDF4 LOC105378948 TNFRSF18 PERM1 C1orf159 TTLL10-AS1 MIR200A KLHL17 NOC2L FAM132A LINC00115 LINC01128 AGRN TNFRSF4 PLEKHN1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR200B LINC01342 TTLL10 TAS1R3 MIR429 TMEM88B FNDC10 LOC107985729 UBE2J2 ATAD3B ATAD3C SSU72 CDK11B B3GALT6 CCNL2 CPSF3L AURKAIP1 ACAP3 SDF4 MIR6808 MXRA8 TMEM240 TNFRSF18 MIR6726 LOC102724312 MMP23B ANKRD65 LOC148413 C1orf159 DVL1 TTLL10-AS1 MIR200A MIR6727 SCNN1D FAM132A ATAD3A CPTP VWA1 TNFRSF4 MRPL20 MIB2 PUSL1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC284600 MIR200B ISG15 LINC01342 TTLL10 TAS1R3 RNF223 MIR429 LOC100288175 TMEM88B UBE2J2 HES4 B3GALT6 CCNL2 SAMD11 CPSF3L LOC100130417 AURKAIP1 ACAP3 LOC107985728 SDF4 LOC105378948 MIR6808 MXRA8 TNFRSF18 MIR6726 PERM1 ANKRD65 LOC148413 C1orf159 DVL1 TTLL10-AS1 MIR200A MIR6727 SCNN1D KLHL17 NOC2L FAM132A CPTP AGRN TNFRSF4 PLEKHN1 MRPL20 PUSL1
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR200B TNFRSF18 LINC01342 TTLL10 RNF223 MIR429 C1orf159 TTLL10-AS1 MIR200A UBE2J2 SCNN1D FAM132A B3GALT6 TNFRSF4 SDF4 LOC105378948
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR200B TNFRSF18 LINC01342 TTLL10 MIR6726 RNF223 MIR429 C1orf159 TTLL10-AS1 MIR200A UBE2J2 SCNN1D FAM132A B3GALT6 TNFRSF4 ACAP3 SDF4
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR200B TNFRSF18 TTLL10 MIR6726 MIR429 TTLL10-AS1 MIR200A MIR6727 UBE2J2 SCNN1D FAM132A B3GALT6 CPSF3L TNFRSF4 ACAP3 SDF4 PUSL1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline