This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07477593_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1261323 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07477593_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss SCNN1D MIR6726 ACAP3 UBE2J2
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TTLL10-AS1 ACAP3 TNFRSF18 MIR200A AGRN SDF4 TAS1R3 MIR200B TNFRSF4 MIR6726 LOC100288175 MIR6727 MIR429 C1orf159 RNF223 TTLL10 CPSF3L LINC01342 CPTP LOC105378948 SCNN1D FAM132A UBE2J2 B3GALT6 PUSL1
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ATAD3B TTLL10-AS1 ATAD3C FAM41C ANKRD65 LOC102724312 SSU72 TMEM240 AGRN LOC100288069 TAS1R3 MIR200B LINC01128 LOC100130417 NADK PLEKHN1 LOC100288175 MIR429 C1orf159 TTLL10 CDK11A KLHL17 LOC148413 LOC107985728 ISG15 CDK11B NOC2L MRPL20 MMP23B LOC105378948 FAM132A MMP23A FNDC10 PUSL1 LOC100287934 SLC35E2B VWA1 DVL1 AURKAIP1 ACAP3 TNFRSF18 MIR200A SDF4 HES4 TNFRSF4 MIR6726 TMEM88B MIR6727 SLC35E2 MIB2 PERM1 LOC107985729 RNF223 MXRA8 LINC00115 LOC284600 FAM87B CPSF3L LINC01342 CPTP SCNN1D SAMD11 UBE2J2 MIR6808 B3GALT6 CCNL2 ATAD3A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CFAP74 MORN1 TTLL10-AS1 LOC100132287 ANKRD65 LOC102724312 SSU72 TMEM240 LOC100288069 MIR200B SKI LINC01128 NADK MIR429 TTLL10 CDK11A OR4F5 LOC107985728 MTND1P23 MIR6859-2 CDK11B LOC100134822 FAM138A LOC105378948 MIR1302-2 PUSL1 GABRD OR4F16 AURKAIP1 OR4F29 LOC105378589 TNFRSF18 MIR6726 TMEM88B PERM1 MXRA8 LOC729737 LINC00115 LOC284600 FAM87B CPSF3L LINC01342 SCNN1D SAMD11 LOC102725121 B3GALT6 CCNL2 LOC100129534 MIR6859-1 ATAD3B LOC107984841 ATAD3C PRKCZ FAM41C FAAP20 AGRN WASH7P TAS1R3 LOC100130417 LOC105378592 PLEKHN1 LOC100288175 C1orf159 LOC105378591 KLHL17 LOC148413 ISG15 NOC2L MRPL20 MMP23B FAM132A MMP23A FNDC10 CALML6 LOC100287934 SLC35E2B TMEM52 VWA1 DDX11L1 DVL1 ACAP3 MIR200A SDF4 HES4 TNFRSF4 LOC105378949 LOC100133331 MIR6727 GNB1 MIR6723 MTND2P28 SLC35E2 MIB2 LOC107985729 RNF223 LOC105378947 CPTP UBE2J2 MIR6808 LOC100506504 LOC101928626 ATAD3A
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss SCNN1D MIR6726 FAM132A ACAP3 UBE2J2 B3GALT6 SDF4
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss TTLL10-AS1 FAM41C TNFRSF18 MIR200A AGRN SDF4 HES4 MIR200B TNFRSF4 LINC01128 LOC100130417 PLEKHN1 LOC100288175 MIR429 C1orf159 PERM1 RNF223 TTLL10 KLHL17 LOC107985728 LINC00115 ISG15 LOC284600 FAM87B NOC2L LINC01342 LOC105378948 SAMD11 FAM132A UBE2J2 B3GALT6
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ATAD3B TTLL10-AS1 VWA1 ATAD3C DVL1 AURKAIP1 ACAP3 ANKRD65 LOC102724312 SSU72 TNFRSF18 MIR200A TMEM240 SDF4 TAS1R3 MIR200B TNFRSF4 MIR6726 TMEM88B MIR6727 MIR429 MIB2 C1orf159 LOC107985729 MXRA8 TTLL10 LOC148413 CDK11B MRPL20 CPSF3L LINC01342 MMP23B CPTP SCNN1D FAM132A UBE2J2 MIR6808 B3GALT6 FNDC10 CCNL2 PUSL1 ATAD3A
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TTLL10-AS1 DVL1 AURKAIP1 ACAP3 ANKRD65 TNFRSF18 MIR200A AGRN SDF4 HES4 TAS1R3 MIR200B TNFRSF4 LOC100130417 MIR6726 PLEKHN1 TMEM88B LOC100288175 MIR6727 MIR429 C1orf159 PERM1 RNF223 MXRA8 TTLL10 KLHL17 LOC148413 LOC107985728 ISG15 LOC284600 NOC2L MRPL20 CPSF3L LINC01342 CPTP LOC105378948 SCNN1D SAMD11 FAM132A UBE2J2 MIR6808 B3GALT6 CCNL2 PUSL1
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10 TTLL10-AS1 TNFRSF18 MIR200A SDF4 LINC01342 MIR200B LOC105378948 TNFRSF4 SCNN1D FAM132A UBE2J2 MIR429 B3GALT6 C1orf159 RNF223
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss TTLL10 TTLL10-AS1 ACAP3 TNFRSF18 MIR200A SDF4 LINC01342 MIR200B TNFRSF4 SCNN1D MIR6726 FAM132A UBE2J2 MIR429 B3GALT6 C1orf159 RNF223
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TTLL10 TTLL10-AS1 ACAP3 TNFRSF18 MIR200A SDF4 CPSF3L MIR200B TNFRSF4 SCNN1D MIR6726 FAM132A UBE2J2 MIR6727 MIR429 B3GALT6 PUSL1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline