This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07479385_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1261423 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07479385_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 UBE2J2 MIR6726 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication ACAP3 C1orf159 AGRN TAS1R3 LOC100288175 MIR200B TNFRSF18 B3GALT6 MIR6727 SCNN1D FAM132A TTLL10-AS1 TTLL10 PUSL1 LOC105378948 CPSF3L MIR6726 MIR429 SDF4 LINC01342 RNF223 UBE2J2 TNFRSF4 CPTP MIR200A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ACAP3 HES4 AGRN LOC107985728 LOC100288175 MMP23B CDK11A SLC35E2 B3GALT6 NADK MIR6727 SCNN1D FAM132A KLHL17 SSU72 SAMD11 LOC105378948 CPSF3L MIR6726 MIR429 LINC00115 CCNL2 LINC01128 AURKAIP1 FNDC10 LOC102724312 ISG15 NOC2L MRPL20 TNFRSF4 CPTP MIB2 LOC107985729 ANKRD65 ATAD3A TMEM240 C1orf159 LOC284600 TAS1R3 MXRA8 MIR200B TNFRSF18 PERM1 LOC100288069 TTLL10-AS1 TTLL10 PUSL1 MIR6808 DVL1 MMP23A TMEM88B LOC100287934 VWA1 SDF4 LINC01342 RNF223 ATAD3C PLEKHN1 SLC35E2B FAM87B LOC100130417 CDK11B UBE2J2 ATAD3B FAM41C LOC148413 MIR200A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss HES4 LOC105378949 AGRN LOC100132287 B3GALT6 MIR6727 SCNN1D FAM132A KLHL17 SSU72 SAMD11 LOC100134822 CPSF3L MIR6726 CALML6 LINC00115 GNB1 LINC01128 OR4F29 AURKAIP1 FNDC10 LOC100506504 LOC102724312 MRPL20 MORN1 CPTP ANKRD65 LOC100133331 ATAD3A LOC284600 MIR200B LOC101928626 TNFRSF18 MIR6859-2 FAM138A LOC100287934 PRKCZ CFAP74 VWA1 MIR6859-1 LINC01342 LOC107984841 RNF223 LOC100130417 CDK11B UBE2J2 FAM41C LOC148413 GABRD LOC105378947 MTND2P28 ACAP3 LOC105378591 LOC107985728 OR4F5 OR4F16 LOC100288175 MMP23B CDK11A LOC105378589 SLC35E2 NADK LOC105378948 MIR6723 MIR429 CCNL2 LOC102725121 DDX11L1 LOC100129534 ISG15 NOC2L TNFRSF4 MIB2 LOC107985729 TMEM240 C1orf159 WASH7P TAS1R3 LOC729737 MXRA8 FAAP20 MTND1P23 PERM1 LOC100288069 TTLL10-AS1 TTLL10 PUSL1 MIR6808 DVL1 MMP23A TMEM88B SDF4 ATAD3C PLEKHN1 SLC35E2B FAM87B LOC105378592 ATAD3B MIR1302-2 SKI TMEM52 MIR200A
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss ACAP3 UBE2J2 B3GALT6 MIR6726 SCNN1D FAM132A SDF4
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss HES4 C1orf159 LOC284600 AGRN LOC107985728 LOC100288175 MIR200B TNFRSF18 B3GALT6 FAM132A PERM1 TTLL10-AS1 KLHL17 TTLL10 SAMD11 LOC105378948 MIR429 SDF4 LINC01342 LINC00115 LINC01128 RNF223 PLEKHN1 FAM87B LOC100130417 UBE2J2 ISG15 FAM41C NOC2L TNFRSF4 MIR200A
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion LOC107985729 ANKRD65 ATAD3A ACAP3 TMEM240 C1orf159 TAS1R3 MXRA8 MMP23B MIR200B TNFRSF18 B3GALT6 MIR6727 SCNN1D FAM132A TTLL10-AS1 SSU72 TTLL10 PUSL1 MIR6808 DVL1 CPSF3L TMEM88B MIR6726 VWA1 MIR429 SDF4 LINC01342 CCNL2 AURKAIP1 ATAD3C CDK11B UBE2J2 FNDC10 ATAD3B LOC102724312 LOC148413 MRPL20 TNFRSF4 CPTP MIR200A MIB2
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain ANKRD65 ACAP3 HES4 C1orf159 LOC284600 AGRN TAS1R3 LOC107985728 MXRA8 LOC100288175 MIR200B TNFRSF18 B3GALT6 MIR6727 SCNN1D FAM132A PERM1 TTLL10-AS1 KLHL17 TTLL10 PUSL1 MIR6808 DVL1 SAMD11 LOC105378948 CPSF3L TMEM88B MIR6726 MIR429 SDF4 LINC01342 CCNL2 RNF223 AURKAIP1 PLEKHN1 LOC100130417 UBE2J2 ISG15 LOC148413 NOC2L MRPL20 TNFRSF4 CPTP MIR200A
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain LOC105378948 C1orf159 MIR429 SDF4 LINC01342 RNF223 MIR200B UBE2J2 TNFRSF18 B3GALT6 SCNN1D FAM132A TTLL10-AS1 TNFRSF4 TTLL10 MIR200A
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss ACAP3 MIR6726 C1orf159 MIR429 SDF4 LINC01342 RNF223 MIR200B UBE2J2 TNFRSF18 B3GALT6 SCNN1D FAM132A TTLL10-AS1 TNFRSF4 TTLL10 MIR200A
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L ACAP3 MIR6726 MIR429 SDF4 MIR200B UBE2J2 TNFRSF18 B3GALT6 MIR6727 SCNN1D FAM132A TTLL10-AS1 TNFRSF4 TTLL10 MIR200A PUSL1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline