This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07480167_cn
See other CPTP CNV Assays ›
Gene Symbol
CPTP
Assay Reference Genome
Location

Chr.1:1327342 on build GRCh38
Cytoband
1p36.33
Assay ID Hs07480167_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

80772

Gene Name:

ceramide-1-phosphate transfer protein

Gene Aliases:

GLTPD1

Location:

Chr.1:1324756-1328896 on Build GRCh38

Assay Gene Location:

Within Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPTP NM_001029885.1 3 639 NP_001025056.1
XM_005244801.3 3 752 XP_005244858.1
XM_005244802.1 2 449 XP_005244859.1
XM_011542200.2 3 648 XP_011540502.1
BC098429.1 1 63 AAH98429.1
JN542538.1 2 224

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PRKCZ MORN1 C1orf159 SLC35E2 MIR429 FAM87B LOC100506504 SSU72 LOC100133331 CFAP74 LOC100288069 RNF223 LOC100132287 CPSF3L MIB2 MIR6726 FAM138A PUSL1 FAM41C LOC105378592 MIR200B LOC107985728 LINC01342 ATAD3C MMP23A AGRN DVL1 TNFRSF4 MIR1302-2 ATAD3B MIR6859-2 KLHL17 CDK11A LOC102725121 TTLL10 OR4F5 LINC01128 WASH7P CDK11B MIR6859-1 FAM132A UBE2J2 LOC105378947 SAMD11 LOC100134822 LOC105378589 OR4F16 DDX11L1 LOC100129534 ANKRD65 LOC107985729 HES4 VWA1 MIR200A LOC105378591 GNB1 TAS1R3 SCNN1D LOC105378949 MXRA8 MMP23B LINC00115 FAAP20 LOC100287934 LOC102724312 OR4F29 CALML6 LOC107984841 TMEM88B AURKAIP1 GABRD SKI LOC284600 TMEM52 MRPL20 CCNL2 MTND1P23 LOC148413 ACAP3 CPTP LOC100288175 LOC729737 PLEKHN1 NADK ISG15 TNFRSF18 SLC35E2B LOC105378948 PERM1 LOC100130417 MIR6808 NOC2L MIR6727 MIR6723 TMEM240 MTND2P28 FNDC10 LOC101928626 TTLL10-AS1 SDF4 B3GALT6 ATAD3A
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss TAS1R3 MIR6808 PUSL1 DVL1 MIR6727 CPSF3L ACAP3 CPTP MIR6726
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss TAS1R3 SCNN1D MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 AURKAIP1 CPTP MIR6726
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain TAS1R3 MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 AURKAIP1 CPTP
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss PUSL1 FAM132A UBE2J2 ACAP3 CPTP C1orf159 MIR200B LINC01342 MIR429 TNFRSF18 MIR200A TAS1R3 SCNN1D MIR6808 DVL1 TNFRSF4 MIR6727 MXRA8 TTLL10-AS1 SDF4 RNF223 TTLL10 CPSF3L B3GALT6 AURKAIP1 MIR6726
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss TAS1R3 MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 CPTP
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPSF3L CPTP
nsv544972 Chr.1:1321172 - 1337736 on Build GRCh38 Loss TAS1R3 DVL1 CPSF3L CPTP
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion TAS1R3 SCNN1D MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 AURKAIP1 CPTP MIR6726
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication PUSL1 FAM132A UBE2J2 ACAP3 CPTP C1orf159 LOC100288175 MIR200B LINC01342 MIR429 TNFRSF18 LOC105378948 MIR200A TAS1R3 SCNN1D AGRN TNFRSF4 MIR6727 TTLL10-AS1 SDF4 RNF223 TTLL10 CPSF3L B3GALT6 MIR6726
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FAM132A UBE2J2 SAMD11 C1orf159 SLC35E2 MIR429 ANKRD65 FAM87B LOC107985729 HES4 VWA1 SSU72 MIR200A TAS1R3 SCNN1D MXRA8 LOC100288069 RNF223 MMP23B LINC00115 LOC100287934 LOC102724312 CPSF3L TMEM88B AURKAIP1 MIB2 MIR6726 LOC284600 PUSL1 MRPL20 CCNL2 FAM41C LOC148413 ACAP3 CPTP LOC100288175 MIR200B PLEKHN1 LOC107985728 LINC01342 NADK ISG15 TNFRSF18 ATAD3C MMP23A SLC35E2B LOC105378948 PERM1 LOC100130417 MIR6808 AGRN DVL1 NOC2L TNFRSF4 MIR6727 ATAD3B TMEM240 FNDC10 KLHL17 TTLL10-AS1 CDK11A SDF4 TTLL10 B3GALT6 ATAD3A LINC01128 CDK11B
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss TAS1R3 MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 CPTP
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain TAS1R3 SCNN1D MIR6808 PUSL1 DVL1 MIR6727 MXRA8 CPSF3L ACAP3 CPTP MIR6726
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion PUSL1 FAM132A MRPL20 CCNL2 UBE2J2 LOC148413 ACAP3 CPTP C1orf159 MIR200B LINC01342 MIR429 TNFRSF18 ATAD3C ANKRD65 LOC107985729 VWA1 SSU72 MIR200A TAS1R3 SCNN1D MIR6808 DVL1 TNFRSF4 MIR6727 ATAD3B TMEM240 MXRA8 FNDC10 TTLL10-AS1 SDF4 MMP23B LOC102724312 TTLL10 CPSF3L B3GALT6 TMEM88B ATAD3A AURKAIP1 MIB2 MIR6726 CDK11B
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss TAS1R3 SCNN1D MIR6808 PUSL1 DVL1 UBE2J2 MIR6727 MXRA8 ACAP3 CPTP CPSF3L AURKAIP1 MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC284600 PUSL1 FAM132A MRPL20 CCNL2 UBE2J2 LOC148413 SAMD11 ACAP3 CPTP C1orf159 LOC100288175 MIR200B PLEKHN1 LOC107985728 LINC01342 ISG15 MIR429 TNFRSF18 ANKRD65 LOC105378948 HES4 PERM1 MIR200A LOC100130417 TAS1R3 SCNN1D MIR6808 AGRN DVL1 NOC2L TNFRSF4 MIR6727 MXRA8 KLHL17 TTLL10-AS1 SDF4 RNF223 TTLL10 CPSF3L B3GALT6 TMEM88B AURKAIP1 MIR6726

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0