This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07490294_cn
See other ADGRE4P CNV Assays ›
Gene Symbol
ADGRE4P
Assay Reference Genome
Location

Chr.19:6953727 on build GRCh38
Cytoband
19p13.2
Assay ID Hs07490294_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

326342

Gene Name:

adhesion G protein-coupled receptor E4, pseudogene

Gene Aliases:

EMR4, EMR4P, FIRE, GPR127, PGR16

Location:

Chr.19:6952500-6990846 on Build GRCh38

Assay Gene Location:

Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ADGRE4P NR_024075.1
AF489700.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762977 Chr.19:6890139 - 7108052 on Build GRCh38 Gain+Loss MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P
esv2751810 Chr.19:6951556 - 7083618 on Build GRCh38 Loss MBD3L4 LOC729458 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P
nsv1061916 Chr.19:6940751 - 7085469 on Build GRCh38 Gain MBD3L4 LOC729458 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P
nsv1056442 Chr.19:6890127 - 6982686 on Build GRCh38 Gain ADGRE1 ADGRE4P
esv3583219 Chr.19:6951972 - 7022131 on Build GRCh38 Loss LOC729458 FLJ25758 ADGRE4P
dgv3421n100 Chr.19:6869658 - 7138691 on Build GRCh38 Gain MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P INSR
esv3574297 Chr.19:6890652 - 7103957 on Build GRCh38 Gain MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P
nsv1056194 Chr.19:6906839 - 7022132 on Build GRCh38 Gain LOC729458 ADGRE1 FLJ25758 ADGRE4P
dgv136e55 Chr.19:6820940 - 7112259 on Build GRCh38 Gain MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 VAV1 MBD3L3 FLJ25758 ZNF557 ADGRE4P INSR
dgv14e196 Chr.19:6710054 - 6986820 on Build GRCh38 Duplication C3 SH2D3A ADGRE1 MIR6791 GPR108 VAV1 TRIP10 ADGRE4P
esv33729 Chr.19:6898574 - 7109534 on Build GRCh38 Gain+Loss MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P
dgv393n27 Chr.19:6895855 - 7112582 on Build GRCh38 Gain MBD3L4 LOC729458 ADGRE1 MBD3L2 MBD3L5 MBD3L3 FLJ25758 ZNF557 ADGRE4P INSR
nsv833729 Chr.19:6832389 - 6977151 on Build GRCh38 Loss ADGRE1 VAV1 ADGRE4P
esv3643571 Chr.19:6896401 - 7019801 on Build GRCh38 Gain LOC729458 ADGRE1 FLJ25758 ADGRE4P
nsv1063661 Chr.19:6942279 - 6982686 on Build GRCh38 Gain ADGRE4P
esv2751809 Chr.19:6896369 - 7816144 on Build GRCh38 Gain MCOLN1 LOC729458 RETN CLEC4GP1 CD209 FCER2 C19orf45 CLEC4G MBD3L4 MCEMP1 MBD3L5 ZNF557 MIR6792 PNPLA6 PEX11G ADGRE1 TRAPPC5 PET100 STXBP2 MBD3L3 FLJ25758 LOC100128573 PCP2 ADGRE4P INSR ZNF358 LOC107985283 MBD3L2 CLEC4M XAB2 CAMSAP3 ARHGEF18

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-695c48667f-9htq6:80/100.66.77.160:80.
git-commit: c086736f0d622c41bbf3ea4d71b1c2b8619a5b9d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.35.0-Offline