This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07534536_cn
See other SALL3 CNV Assays ›
Gene Symbol
SALL3
Assay Reference Genome
Location

Chr.18:78980340 on build GRCh38
Cytoband
18q23
Assay ID Hs07534536_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

27164

Gene Name:

spalt like transcription factor 3

Gene Aliases:

ZNF796

Location:

Chr.18:78980275-78998969 on Build GRCh38

Assay Gene Location:

Overlaps Exon 1 - Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SALL3 NM_171999.3 NP_741996.2
BC148296.2

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1071874 Chr.18:78978999 - 78980600 on Build GRCh38 Deletion SALL3
nsv577821 Chr.18:78980085 - 78994367 on Build GRCh38 Loss SALL3
nsv1133432 Chr.18:78976800 - 78980600 on Build GRCh38 Deletion SALL3
nsv1143887 Chr.18:78978700 - 78980600 on Build GRCh38 Deletion SALL3
nsv577793 Chr.18:78951024 - 79022312 on Build GRCh38 Loss SALL3
esv2666555 Chr.18:78977945 - 78981392 on Build GRCh38 Deletion SALL3
nsv577795 Chr.18:78972221 - 79050030 on Build GRCh38 Loss SALL3
nsv577796 Chr.18:78978993 - 78980500 on Build GRCh38 Loss SALL3
dgv564n67 Chr.18:78952775 - 79013936 on Build GRCh38 Gain SALL3
nsv828342 Chr.18:78976549 - 78983544 on Build GRCh38 Gain SALL3
dgv6103n54 Chr.18:78979851 - 78980976 on Build GRCh38 Loss SALL3
nsv828338 Chr.18:78963814 - 79001729 on Build GRCh38 Gain SALL3
dgv6104n54 Chr.18:78980034 - 78980976 on Build GRCh38 Gain+Loss SALL3
esv2717497 Chr.18:78170182 - 79635788 on Build GRCh38 Deletion NFATC1 SALL3 LOC284240 LOC107985149 ATP9B LOC100996274
nsv1141945 Chr.18:78170394 - 79635820 on Build GRCh38 Deletion NFATC1 SALL3 LOC284240 LOC107985149 ATP9B LOC100996274
dgv565n67 Chr.18:78974294 - 78993921 on Build GRCh38 Gain SALL3
dgv6101n54 Chr.18:78979457 - 78980923 on Build GRCh38 Gain SALL3
nsv828333 Chr.18:78955949 - 79017302 on Build GRCh38 Loss SALL3
nsv577794 Chr.18:78951366 - 79472709 on Build GRCh38 Gain NFATC1 SALL3 LOC107985149 ATP9B LOC100996274
esv29012 Chr.18:78979427 - 78981039 on Build GRCh38 Loss SALL3
nsv1065902 Chr.18:78538045 - 80257174 on Build GRCh38 Gain PARD6G-AS1 LOC105376875 TXNL4A SALL3 RBFADN ADNP2 HSBP1L1 PARD6G LOC284240 LOC107985149 CTDP1 NFATC1 PQLC1 LOC284241 KCNG2 ATP9B RBFA LOC100996274
esv3643277 Chr.18:78784238 - 78988253 on Build GRCh38 Loss SALL3

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
olfactory bulb interneuron development
neurogenesis
forelimb morphogenesis
hindlimb morphogenesis
negative regulation of smoothened signaling pathway
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0