This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07539564_cn
See other CHL1 CNV Assays ›
Gene Symbol
CHL1
Assay Reference Genome
Location

Chr.3:273239 on build GRCh38
Cytoband
3p26.3
Assay ID Hs07539564_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10752

Gene Name:

cell adhesion molecule L1 like

Gene Aliases:

CALL, L1CAM2

Location:

Chr.3:196588-409417 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHL1 NM_001253387.1 NP_001240316.1
NM_006614.3 NP_006605.2
XM_006712938.1 XP_006713001.1
XM_006712939.3 XP_006713002.1
XM_006712940.3 XP_006713003.1
XM_011533292.1 XP_011531594.1
XM_011533294.1 XP_011531596.1
XM_011533295.1 XP_011531597.1
XM_011533296.1 XP_011531598.1
XM_017005566.1 XP_016861055.1
XM_017005567.1 XP_016861056.1
XM_017005569.1 XP_016861058.1
XM_017005570.1 XP_016861059.1
XM_017005571.1 XP_016861060.1
XM_017005572.1 XP_016861061.1
XM_017005573.1 XP_016861062.1
AB209329.1 BAD92566.1
AF002246.1 AAB60937.1
AK126878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2759122 Chr.3:29482 - 756820 on Build GRCh38 Gain+Loss CHL1 LINC01266 LOC101927174 CHL1-AS1
esv3594873 Chr.3:36658 - 319672 on Build GRCh38 Gain CHL1 LOC101927174
nsv818121 Chr.3:272855 - 283727 on Build GRCh38 Gain CHL1
esv2667629 Chr.3:39576 - 1039187 on Build GRCh38 Deletion CHL1 LINC01266 LOC101927174 CHL1-AS1 CNTN6
nsv1013825 Chr.3:256421 - 312812 on Build GRCh38 Loss CHL1
esv3575528 Chr.3:241068 - 275532 on Build GRCh38 Gain CHL1
dgv4608n100 Chr.3:18655 - 438490 on Build GRCh38 Gain CHL1 LOC101927174 CHL1-AS1 LOC102723448
nsv834589 Chr.3:235505 - 395503 on Build GRCh38 Gain CHL1 CHL1-AS1
dgv4613n100 Chr.3:52474 - 1799230 on Build GRCh38 Loss CHL1 LINC01266 LOC101927174 CHL1-AS1 CNTN6
nsv589288 Chr.3:228838 - 275462 on Build GRCh38 Loss CHL1
nsv10238 Chr.3:271826 - 285163 on Build GRCh38 Gain CHL1
dgv4614n100 Chr.3:214575 - 294771 on Build GRCh38 Gain CHL1
nsv1010970 Chr.3:188327 - 1623352 on Build GRCh38 Gain CHL1 LINC01266 LOC101927174 CHL1-AS1 CNTN6
nsv469898 Chr.3:21733 - 886773 on Build GRCh38 Loss CHL1 LINC01266 LOC101927174 CHL1-AS1 LOC102723448
esv3594894 Chr.3:242814 - 379641 on Build GRCh38 Gain CHL1 CHL1-AS1
nsv428412 Chr.3:29467 - 326726 on Build GRCh38 Gain CHL1 LOC101927174
nsv589283 Chr.3:192626 - 321571 on Build GRCh38 Gain CHL1 LOC101927174
nsv999459 Chr.3:230573 - 288287 on Build GRCh38 Loss CHL1
nsv1005290 Chr.3:247092 - 949319 on Build GRCh38 Loss CHL1 LINC01266 CHL1-AS1
dgv4615n100 Chr.3:243604 - 278294 on Build GRCh38 Gain CHL1
esv24432 Chr.3:271980 - 284867 on Build GRCh38 Gain CHL1
nsv1004600 Chr.3:244100 - 607771 on Build GRCh38 Loss CHL1 LINC01266 CHL1-AS1
dgv8152n54 Chr.3:245736 - 278518 on Build GRCh38 Loss CHL1
esv3893589 Chr.3:251309 - 275462 on Build GRCh38 Gain CHL1
esv3893591 Chr.3:264957 - 301339 on Build GRCh38 Loss CHL1
nsv589242 Chr.3:47730 - 388999 on Build GRCh38 Gain CHL1 LOC101927174 CHL1-AS1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell adhesion
signal transduction
axon guidance
adult locomotory behavior
axon regeneration
exploration behavior
negative regulation of neuron apoptotic process
cognition
protease binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-548c9458fd-zf69l:80/100.66.73.23:80.
git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.0-Offline