This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00013169_cn
See other HSPD1 CNV Assays ›
Gene Symbol
Hspd1
Assay Reference Genome
Location

Chr.1:55083073 on build GRCm38
Cytoband
1C1.2
Assay ID Mm00013169_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

15510

Gene Name:

heat shock protein 1 (chaperonin)

Gene Aliases:

60kDa, Hsp60

Location:

Chr.1:55077834-55088112 on Build GRCm38

Assay Gene Location:

Overlaps Intron 5 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Hspd1 NM_010477.4 NP_034607.3
AK088844.1 BAC40607.1
AK143882.1 BAE25581.1
AK146831.1 BAE27466.1
AK177219.1
AK179525.1
AK184625.1
AK187612.1
AK188312.1
AK216069.1
AK217800.1
BC016400.1 AAH16400.1
BC106112.1 AAI06113.1
X53584.1 CAA37653.1
X55023.1 CAA38762.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic

Gene Ontology Categories:

Function(s) Process(es)

detection of misfolded protein
B cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
positive regulation of T cell mediated immune response to tumor cell
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
positive regulation of interferon-alpha production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
protein refolding
B cell proliferation
T cell activation
B cell activation
positive regulation of macrophage activation
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
isotype switching to IgG isotypes
positive regulation of inflammatory response
protein stabilization
positive regulation of T cell activation
chaperone mediated protein folding requiring cofactor
nucleotide binding
lipopolysaccharide binding
protease binding
p53 binding
double-stranded RNA binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
insulin binding
poly(A) RNA binding
protein heterodimerization activity
chaperone binding
misfolded protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline