This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00031169_cn
See other NDUFS1 CNV Assays ›
Gene Symbol
Ndufs1
Assay Reference Genome
Location

Chr.1:63159718 on build GRCm38
Cytoband
1C2
Assay ID Mm00031169_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

227197

Gene Name:

NADH dehydrogenase (ubiquinone) Fe-S protein 1

Gene Aliases:

5830412M15Rik, 9930026A05Rik

Location:

Chr.1:63143592-63176822 on Build GRCm38

Assay Gene Location:

Overlaps Intron 11 - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Ndufs1 NM_001160038.1 NP_001153510.1
NM_001160039.1 NP_001153511.1
NM_001160040.1 NP_001153512.1
NM_145518.2 NP_663493.2
AK034597.1 BAE20494.1
AK036926.1 BAC29641.1
AK078590.1 10 1108
AK142711.1 BAE25170.1
AK146814.1 BAE27454.1
AK166604.1
AK167705.1 BAE39749.1
BC006660.1 AAH06660.1
BC015300.1 AAH15300.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

transport
apoptotic mitochondrial changes
ATP synthesis coupled electron transport
cellular respiration
ATP metabolic process
regulation of mitochondrial membrane potential
oxidation-reduction process
reactive oxygen species metabolic process
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0