This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00042419_cn
See other AK8 CNV Assays ›
Gene Symbol
Ak8
Assay Reference Genome
Location

Chr.2:28699532 on build GRCm38
Cytoband
2A3
Assay ID Mm00042419_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

68870

Gene Name:

adenylate kinase 8

Gene Aliases:

1190002A17Rik, AK 8

Location:

Chr.2:28698226-28813165 on Build GRCm38

Assay Gene Location:

Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Ak8

Back To Top

More Information


Set Membership:

Intragenic Non-exonic

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-vvlg5:80/100.66.78.195:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline