This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00098837_cn
See other NUP210L CNV Assays ›
Gene Symbol
Nup210l
Assay Reference Genome
Location

Chr.3:90129148 on build GRCm38
Cytoband
3F1
Assay ID Mm00098837_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

77595

Gene Name:

nucleoporin 210-like

Gene Aliases:

4930548O11Rik, R26-EGFP, Tg(Gt(ROSA)26Sor-EGFP)130910Eps

Location:

Chr.3:90101569-90212151 on Build GRCm38

Assay Gene Location:

Overlaps Intron 7 - Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Nup210l NM_029937.1 NP_084213.1
AK019752.1 BAB31846.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
Sertoli cell development
molecular_function

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline