Assay Details
Target Gene Details
Entrez Gene ID: | 77595 |
Gene Name: | nucleoporin 210-like |
Gene Aliases: |
4930548O11Rik, R26-EGFP, Tg(Gt(ROSA)26Sor-EGFP)130910Eps |
Location: |
Chr.3:90101569-90212151 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup210l | NM_029937.1 | NP_084213.1 | ||
| AK019752.1 | BAB31846.1 |
More Information
Set Membership: |
|

Genomic Map