This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00113063_cn
See other RSG1 CNV Assays ›
Gene Symbol
Rsg1
Assay Reference Genome
Location

Chr.4:141218608 on build GRCm38
Cytoband
4D3
Assay ID Mm00113063_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

76166

Gene Name:

REM2 and RAB-like small GTPase 1

Gene Aliases:

6330545A04Rik, Gm723

Location:

Chr.4:141213666-141220365 on Build GRCm38

Assay Gene Location:

Within Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Rsg1 NM_001081174.2 5 607 NP_001074643.1

Back To Top

More Information


Set Membership:

Intragenic Exonic

Gene Ontology Categories:

Function(s) Process(es)

transport
exocytosis
small GTPase mediated signal transduction
protein transport
cell projection organization
nucleotide binding
GTP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline