Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag2 | NM_001170555.1 | NP_001164026.1 | ||
| NM_001170556.1 | NP_001164027.1 | |||
| NM_145401.2 | NP_663376.2 | |||
| AK010703.1 | ||||
| AK032238.1 | BAC27775.1 | |||
| AK154296.1 | BAE32497.1 | |||
| AK165834.1 | BAE38403.1 | |||
| AY348864.1 | AAQ55224.1 | |||
| BC015283.1 | AAH15283.1 | |||
| BQ572978.1 | ||||
| BU707674.1 |
More Information
Set Membership: |
|

Genomic Map