Assay Details
Target Gene Details
Entrez Gene ID: | 19053 |
Gene Name: | protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform |
Gene Aliases: |
AI115466, D8Ertd766e, PP2Ac |
Location: |
Chr.8:33599621-33619804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2cb | NM_017374.3 | NP_059070.1 | ||
| AK090311.1 | BAC41164.1 | |||
| BC019161.1 | AAH19161.1 | |||
| BC058582.1 | AAH58582.1 | |||
| BE847789.1 | ||||
| Z67746.1 | CAA91559.1 |
More Information
Set Membership: |
|

Genomic Map