Assay Details
Target Gene Details
Entrez Gene ID: | 11933 |
Gene Name: | ATPase, Na+/K+ transporting, beta 3 polypeptide |
Gene Aliases: |
AA409958, AI664000, AW212096 |
Location: |
Chr.9:96332662-96364905 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1b3 | NM_007502.4 | NP_031528.1 | ||
| AK032516.1 | BAC27906.1 | |||
| AK148495.1 | BAE28587.1 | |||
| AK151700.1 | BAE30623.1 | |||
| AK151841.1 | BAE30733.1 | |||
| AK159532.1 | BAE35160.1 | |||
| AK161282.1 | ||||
| AK168273.1 | BAE40219.1 | |||
| AK177007.1 | ||||
| AK218119.1 | ||||
| BC079916.1 | ||||
| U59761.1 | AAC00019.1 |
More Information
Set Membership: |
|

Genomic Map