Assay Details
Target Gene Details
Entrez Gene ID: | 216739 |
Gene Name: | acyl-CoA synthetase long-chain family member 6 |
Gene Aliases: |
A330035H04Rik, AW050338, Facl6, LACS, Lacsl, mKIAA0837 |
Location: |
Chr.11:54303824-54364756 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsl6 | NM_001033597.1 | 2 | 233 | NP_001028769.1 |
| NM_001033598.1 | 2 | 202 | NP_001028770.1 | |
| NM_001033599.1 | 2 | 408 | NP_001028771.1 | |
| NM_144823.4 | 2 | 233 | NP_659072.3 | |
| AB073974.1 | 1 | 77 | ||
| AK032479.1 | 3 | 492 | BAC27892.1 | |
| AK039380.1 | 2 | 319 | BAC30332.1 | |
| AK147283.1 | 2 | 203 | BAE27819.1 | |
| AK147421.1 | 2 | 234 | BAE27902.1 | |
| AY167035.1 | 2 | 203 | AAO38689.1 | |
| AY786360.1 | 4 | 751 | AAW33883.1 | |
| AY786361.1 | 2 | 183 | AAW33884.1 | |
| AY786362.1 | 2 | 375 | AAW33885.1 | |
| AY786363.1 | 2 | 228 | AAW33886.1 | |
| BB634066.1 | 2 | 408 | ||
| BC016114.1 | 2 | 159 | AAH16114.1 | |
| BC022959.1 | 2 | 233 |
More Information
Set Membership: |
|

Genomic Map