Assay Details
Target Gene Details
Entrez Gene ID: | 380702 |
Gene Name: | shisa family member 6 |
Gene Aliases: |
Gm879 |
Location: |
Chr.11:66211725-66526126 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shisa6 | NM_001034874.3 | 4 | 1118 | NP_001030046.1 |
AK147504.1 | 4 | 1119 | BAE27958.1 |
