Assay Details
Target Gene Details
Entrez Gene ID: | 217351 |
Gene Name: | trinucleotide repeat containing 6C |
Gene Aliases: |
9930033H14Rik, mKIAA1582 |
Location: |
Chr.11:117654289-117763439 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnrc6c | NM_198022.2 | NP_932139.2 | ||
| AK036989.1 | ||||
| AK122529.1 | BAC65811.1 | |||
| AK147471.1 | BAE27937.1 | |||
| AK147551.1 |
More Information
Set Membership: |
|

Genomic Map