Assay Details
Target Gene Details
Entrez Gene ID: | 218503 |
Gene Name: | FCH domain only 2 |
Gene Aliases: |
5832424M12Rik, AA387320 |
Location: |
Chr.13:98723406-98815769 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcho2 | NM_172591.3 | 21 | 2150 | NP_766179.2 |
| AK031041.1 | 21 | 2153 | BAC27226.1 | |
| AK142282.1 | 21 | 1836 | BAE25007.1 | |
| AK161093.1 | 2 | 883 | BAE36192.1 | |
| AK164135.1 | 22 | 2008 | BAE37642.1 | |
| AK169439.1 | 5 | 1572 | ||
| BC052456.1 | 21 | 1847 | AAH52456.1 |
More Information
Set Membership: |
|

Genomic Map