This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443094_cn
See other FAM3B CNV Assays ›
Gene Symbol
Fam3b
Assay Reference Genome
Location

Chr.16:97475819 on build GRCm38
Cytoband
16C4
Assay ID Mm00443094_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

52793

Gene Name:

family with sequence similarity 3, member B

Gene Aliases:

2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander

Location:

Chr.16:97471086-97504936 on Build GRCm38

Assay Gene Location:

Overlaps Intron 5 - Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Fam3b NM_020622.2 NP_065647.1
AF360358.1 AAL34461.1
AF494380.1 AAM94281.1
AK018571.1 BAB31283.1
BC048949.1 AAH48949.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic

Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
insulin secretion
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0