Assay Details
Target Gene Details
Entrez Gene ID: | 22350 |
Gene Name: | ezrin |
Gene Aliases: |
AW146364, R75297, Vil2, p81 |
Location: |
Chr.17:6738131-6782780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ezr | NM_009510.2 | NP_033536.2 | ||
| AK002766.1 | BAB22341.1 | |||
| AK004588.2 | ||||
| AK035271.1 | BAC29009.1 | |||
| AK135194.1 | BAE22456.1 | |||
| AK135353.1 | BAE22499.1 | |||
| AK145707.1 | BAE26602.1 | |||
| AK159717.1 | BAE35312.1 | |||
| AK159824.1 | BAE35404.1 | |||
| AK159937.1 | ||||
| AK166191.1 | BAE38621.1 | |||
| AK168410.1 | ||||
| AK168840.1 | BAE40663.1 | |||
| AK170610.1 | BAE41910.1 | |||
| AK172336.1 | BAE42954.1 | |||
| AK189604.1 | ||||
| BC048181.1 | AAH48181.2 | |||
| BC098502.1 | AAH98502.1 | |||
| X60671.1 | CAA43086.1 |
More Information
Set Membership: |
|

Genomic Map