Assay Details
Target Gene Details
Entrez Gene ID: | 20359 |
Gene Name: | sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B |
Gene Aliases: |
Sema, Seman, VIb, semaZ |
Location: |
Chr.17:56123085-56140343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema6b | NM_001130456.1 | NP_001123928.1 | ||
| NM_013662.2 | NP_038690.1 | |||
| AF036585.1 | AAC00493.1 | |||
| AK139358.1 | BAE23975.1 | |||
| AK143229.1 | ||||
| AK166077.1 | BAE38558.1 | |||
| BC139415.1 |
More Information
Set Membership: |
|

Genomic Map