This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00493773_cn
See other ESRRA CNV Assays ›
Gene Symbol
Esrra
Assay Reference Genome
Location

Chr.19:6914096 on build GRCm38
Cytoband
19A
Assay ID Mm00493773_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26379

Gene Name:

estrogen related receptor, alpha

Gene Aliases:

ERRalpha, Err1, Estrra, Nr3b1

Location:

Chr.19:6910977-6922048 on Build GRCm38

Assay Gene Location:

Overlaps Intron 3 - Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Esrra NM_007953.2 NP_031979.2
AK019222.1
AK156371.1 BAE33690.1
BC138586.1
BC171958.1
CO425338.1
U85259.1 AAB51250.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
regulation of ossification
regulation of cell proliferation
steroid hormone mediated signaling pathway
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cartilage development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
steroid binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline