This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549610_cn
See other AW011738 CNV Assays ›
Gene Symbol
AW011738
Assay Reference Genome
Location

Chr.4:156204193 on build GRCm38
Cytoband
4E2
Assay ID Mm00549610_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100382

Gene Name:

expressed sequence AW011738

Gene Aliases:

-

Location:

Chr.4:156203284-156206028 on Build GRCm38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AW011738 NR_030671.1 1 910
AK036383.1 1 891
AK078660.1 1 892
AK086145.1 1 911
AK156747.1 1 895 BAE33835.1
AK172062.1 2 725 BAE42805.1
AK172315.1 2 725 BAE42940.1

Target Gene Details

Entrez Gene ID:

74183

Gene Name:

PPARGC1 and ESRR induced regulator, muscle 1

Gene Aliases:

2310042D19Rik

Location:

Chr.4:156203268-156221307 on Build GRCm38

Assay Gene Location:

Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Perm1

Back To Top

More Information


Set Membership:

Intragenic Exonic Non-exonic

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline