This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555112_cn
See other TNFRSF4 CNV Assays ›
Gene Symbol
Tnfrsf4
Assay Reference Genome
Location

Chr.4:156013899 on build GRCm38
Cytoband
4E2
Assay ID Mm00555112_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

22163

Gene Name:

tumor necrosis factor receptor superfamily, member 4

Gene Aliases:

ACT35, CD134, Ly-70, Ox40, TXGP1L, Txgp1

Location:

Chr.4:156013695-156016613 on Build GRCm38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Tnfrsf4 NM_011659.2 1 205 NP_035789.1
BC139239.1 1 75
BC139266.1 1 75
Z21674.1 CAA79772.1

Back To Top

More Information


Set Membership:

Intragenic Exonic

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cellular defense response
programmed cell death
positive regulation of B cell proliferation
tumor necrosis factor-mediated signaling pathway
T cell proliferation
regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
regulation of protein kinase activity
negative regulation of transcription, DNA-templated
negative regulation of cytokine secretion
positive regulation of immunoglobulin secretion
negative regulation of activation-induced cell death of T cells
negative regulation of extrinsic apoptotic signaling pathway
tumor necrosis factor-activated receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0