Assay Details
Target Gene Details
Entrez Gene ID: | 14871 |
Gene Name: | glutathione S-transferase, theta 1 |
Gene Aliases: |
AI255817, Gstt1-1 |
Location: |
Chr.10:75783813-75798584 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstt1 | NM_008185.3 | 5 | 765 | NP_032211.3 |
| AI255817.1 | 1 | 285 | ||
| AK002338.1 | 5 | 765 | BAB22023.1 | |
| BC012254.1 | 5 | 680 | AAH12254.1 | |
| BC055020.1 | 5 | 701 | AAH55020.1 | |
| X98055.1 | 5 | 677 | CAA66665.1 |

Genomic Map