This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00596836_cn
See other NR1D1 CNV Assays ›
Gene Symbol
Nr1d1
Assay Reference Genome
Location

Chr.11:98774976 on build GRCm38
Cytoband
11D
Assay ID Mm00596836_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

217166

Gene Name:

nuclear receptor subfamily 1, group D, member 1

Gene Aliases:

A530070C09Rik, R75201

Location:

Chr.11:98767932-98775377 on Build GRCm38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Nr1d1 NM_145434.4 1 402 NP_663409.2
AF291821.1 1 226 AAG01345.1
AK041047.1 1 257
AK135640.1 1 264
AK137398.1 1 289 BAE23342.1
AK137582.1 1 267 BAE23418.1
AK146430.1 1 359 BAE27164.1
AK154931.1 1 403 BAE32932.1
AK155597.1 1 403 BAE33339.1
X86010.1 1 262 CAA59997.1

Back To Top

More Information


Set Membership:

Intragenic Exonic

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
glycogen biosynthetic process
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
circadian rhythm
proteasomal protein catabolic process
negative regulation of receptor biosynthetic process
regulation of lipid metabolic process
cell differentiation
circadian regulation of gene expression
negative regulation of toll-like receptor 4 signaling pathway
regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
regulation of circadian rhythm
steroid hormone mediated signaling pathway
response to leptin
regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
rhythmic process
circadian temperature homeostasis
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
positive regulation of bile acid biosynthetic process
cellular response to lipopolysaccharide
regulation of cholesterol homeostasis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription corepressor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
protein binding
zinc ion binding
heme binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline